Proteomics and Protein Expr.
Protein Expression
Cloning and Expression
Cell Lysis
Protocols
Purification and Detection
Protein Analysis
Stable Isotopes
Stem Cell Biology
Your Favorite Gene - Search
Life Science Innovations
PathFinder
 Product List for "General Method for MALDI-MS Analysis of Proteins and Peptides" Protocol
Sigma-Aldrich Research Biotechnology
The following Cold Spring Harbor Laboratory Press protocols are brought to you by BioSupplyNet in a partnership with Sigma-Aldrich. A list of products is available to use in each protocol.
View the protocol on BioSupplyNet.com.
Selected Protocol from "Proteins and Proteomics: A Laboratory Manual", Richard J. Simpson.
General Method for MALDI-MS Analysis of Proteins and Peptides
David Frecklington
Joint Proteomics Laboratory (JPSL)
Ludwig Institute for Cancer Research
Walter and Eliza Hall Institute of Medical Research
Melbourne, Australia
ABSTRACT
For MALDI-TOF-MS analysis of proteins and peptides, samples are cocrystallized with an excess of organic matrix that absorbs at a specific wavelength (usually, UV337nm). Typically, sinapinic acid (SA) is the matrix of choice for large proteins, whereas {alpha}-cyano-4-hydroxy-cinnamic acid (HCCA) is the preferred matrix for peptide mapping. Following a short laser pulse, analytes are protonated and desorbed into the gas phase, and their m/z values are determined in a TOF mass analyzer. Mass accuracy determinations vary from ±0.01% to 0.1% depending on the sample preparation technique and the method used for mass calibration.
Buy this book, Z700428
Products Available for this Protocol
Protocol Material Description Product #  Product Name Add to Cart
Buffers, Solutions, and Reagents      
Acetonitrile 34967 Acetonitrile
Methanol M1775 Methanol, Absolute - Acetone free
TFA T6508 Trifluoroacetic acid, reagent grade
n-octyl glucoside O8001 Octyl β-D-glucopyranoside
Standards Peptide and protein standards for mass spectrometry analysis
MALDI-MS matrices Matrices for MALDI Analysis
Visit the Mass Spectrometry Site for all of your Mass Spec needs.
back to top
View all available Cold Spring Harbor Laboratory Press Protocols
Product Association Disclaimer: The Sigma-Aldrich products listed for this specific protocol were selected either to match or to supplement the products listed within the actual protocol. The products/reagents from Sigma-Aldrich have been qualified for usage, but may not have been validated for this specific application. Please refer to the detailed product description on the usage of specific products of interest.