Proteomics and Protein Expr.
Protein Expression
Cloning and Expression
Cell Lysis
Protocols
Purification and Detection
Protein Analysis
Stable Isotopes
Stem Cell Biology
Your Favorite Gene - Search
Life Science Innovations
PathFinder
 Product List for “Characterization of Phosphopeptides Using a Combination
 of Immobilized Metal Ion Affinity Media and Direct Analysis by MALDI-TOF-MS”
 Protocol
Proteomics
The following Cold Spring Harbor Laboratory Press protocols are brought to you by BioSupplyNet in a partnership with Sigma-Aldrich. A list of products is available to use in each protocol.
View the protocol on BioSupplyNet.com.
Selected Protocol from "Proteins and Proteomics: A Laboratory Manual," David R. Goodlett, Eugene C. Yi, and Philippe Mottay.
View the protocol "Characterization of Phosphopeptides Using a Combination of Immobilized Metal Ion Affinity Media and Direct Analysis by MALDI-TOF-MS" at BioSupplynet.com
Leesa J. Deterding, Jenny M. Cutalo, and Kenneth B. Tomer
ABSTRACT
Phosphoproteins and peptides can be bound with high specificity to immobilized metal ions such as Fe3+, Ni2+, and Ga3+. This technique can be used with either on-line or off-line MS analysis. However, elution of the phosphopeptides from the metal ion column prior to MS analysis can result in sample loss. Affinity-bound analytes, including phosphopeptides, can be directly analyzed by MALDI-MS without prior elution from the affinity media. Also, consecutive enzymatic reactions, such as phosphatase or carboxypeptidase Y digestion, can be carried out on affinity-bound peptides. When the affinity-bound phosphopeptides are treated with phosphatase, the number of phosphorylation sites can be determined based on the observation of 80-Da (or multiples of 80 Da) mass shifts in the MALDI-MS of the reaction mixture. Carboxypeptidase Y treatment of the affinity-bound phosphopeptides can also be used to cleave amino acids from the carboxyl terminus, with subsequent direct analysis of the enzymatic products by MALDI-MS to locate the phosphorylation sites on the bound phosphopeptides. This protocol details the preparation and use of Fe3+ or Ga3+ metal IMAC with the on-bead analysis of phosphopeptides by MALDI-MS. Enzymatic digestion of affinity-bound peptides is also described.
Buy this book, Z700428
Products Available for this Protocol
Protocol Material Description Product # Product Name Add to Cart
Acetic acid A6283 Acetic acid, ReagentPlus®, ≥99%
Ammonium bicarbonate A6141 Ammonium bicarbonate, ReagentPlus®, ≥99.0%
Calf intestinal alkaline phosphatase 79385 Phosphatase, Alkaline from bovine intestinal mucosa, BioChemika, solution (clear), >10000 U/mL
EDTA E5134 Ethylenediaminetetraacetic acid disodium salt dihydrate, for molecular biology, ≥99%
Carboxypeptidase Y C3888 Carboxypeptidase Y from baker's yeast (S. cerevisiae), lyophilized powder, ≥50 units/mg protein
Ferric chloride (FeCl3) F7134 Iron(III) chloride
Gallium chloride (GaCl3) 427128 Gallium(III) chloride, anhydrous, beads, -10 mesh, 99.999+ %
Ethanol E7148 Ethanol, for molecular biology, 190 proof
H2O W4502 Water, Molecular Biology Reagent
Formic acid F0507 Formic acid, reagent grade, ≥95%
Sodium citrate S1804 Sodium citrate, meets USP testing specifications
TPCK-modified trypsin (sequence-grade) 93630 Trypsin, TPCK treated, from bovine pancreas, BioChemika, ~7500 units/mg
back to top
View all available Cold Spring Harbor Laboratory Press Protocols
Product Association Disclaimer: The Sigma-Aldrich products listed for this specific protocol were selected either to match or to supplement the products listed within the actual protocol. The products/reagents from Sigma-Aldrich have been qualified for usage, but may not have been validated for this specific application. Please refer to the detailed product description on the usage of specific products of interest.