Proteomics and Protein Expr.
Protein Expression
Cloning and Expression
Cell Lysis
Protocols
Purification and Detection
Protein Analysis
Stable Isotopes
Stem Cell Biology
Your Favorite Gene - Search
Life Science Innovations
PathFinder
 Product List for "Preparation of Reversed-Phase Microcolumns" Protocol
Proteomics
The following Cold Spring Harbor Laboratory Press protocols are brought to you by BioSupplyNet in a partnership with Sigma-Aldrich. A list of products is available to use in each protocol.
View the protocol on BioSupplyNet.com.
View the protocol, "Preparation of Reversed-Phase Microcolumns," at BioSupplynet.com.
Selected protocol from "Proteins and Proteomics: A Laboratory Manual," Richard J. Simpson.
ABSTRACT
One versatile strategy for sample cleanup prior to MALDI-MS analysis uses microscale columns designed for direct sample elution onto the MALDI target plate. This protocol describes the fabrication of a reversed-phase microcolumn designed for this purpose. The microcolumns are prepared from GELoader tips. This protocol has been optimized for sample cleanup prior to MALDI-MS. However, with slight modifications, it works equally well with samples destined for ESI-MS.
Proteins and Proteomics: A Laboratory Manual
Buy this book,
Z700428
Products Available for this Protocol
Protocol Material Description Product # Product Name Add to Cart
Chromatography resins Chromatography Resins Selection
Formic acid 251364 Formic acid, ACS reagent, ≥96.0%
SA (sinapinic acid) S8313 Sinapinic Acid, Use tested for MALDI-TOF Mass Spec.
DHB (2,5-dihydroxybenzoic acid) 85707 2,5-Dihydroxybenzoic acid, puriss. p.a., matrix substance for MALDI-MS, >99.0% (HPLC)
Acetonitrile 34998 Acetonitrile, CHROMASOLV® Plus, for HPLC, ≥99.9%
CHCA (alpha-cyano-4-hydroxy-cinnamic acid) 70990 α-Cyano-4-hydroxycinnamic acid, puriss. p.a., matrix substance for MALDI-MS, ≥99.0%
TFA (trifluoroacetic acid) 302031 Trifluoroacetic acid, spectrophotometric grade, 99+ %
Methanol 34860 Methanol, CHROMASOLV®, for HPLC, ≥99.9%
Back to Top
View all available Cold Spring Harbor Laboratory Press Protocols
Product Association Disclaimer: The Sigma-Aldrich products listed for this specific protocol were selected either to match or to supplement the products listed within the actual protocol. The products/reagents from Sigma-Aldrich have been qualified for usage, but may not have been validated for this specific application. Please refer to the detailed product description on the usage of specific products of interest.