Proteomics and Protein Expr.
Protein Expression
Cloning and Expression
Cell Lysis
Protocols
Purification and Detection
Protein Analysis
Stable Isotopes
Stem Cell Biology
Your Favorite Gene - Search
Life Science Innovations
PathFinder
 EZview™ Red - High Visibility Affinity Gel
Purification & Detection
Technical Poster
Data
Ordering Information
A major disadvantage of affinity-based molecular pull-down and immunoprecipitation procedures is that the affinity matrix is difficult to see in the microcentrifuge tube following centrifugation steps.
To facilitate more speed and accuracy, Sigma has developed a dye-conjugated agarose for our EZview Red Affinity Gels. The vivid red color of the affinity beads provides high visibility that allows easy differentiation of the pellet from the supernatant, therefore reducing the risk of accidental aspiration of the pellet and allowing for less tedious manipulations.
Maximize yields: Clear meniscus definition for improved target protein recovery
Reproducibility: Avoid sample loss through accidental aspiration of the pellet
Convenience: Highly visible agarose give you the speed and confidence
Technical Poster:
Affinity Capture Resins With Enhanced Visibility (80 kb PDF)
Data:
Figure 1. EZview Red Protein A Affinity Gel (left, Product Code: P6486) has enhanced visibility compared to standard, non-colored Protein A Agarose (right, Product Code: P2545) Figure 2. EZview Red Protein Affinity Gel (Product Code: P2545) recovered a similar amount IgG. Different amounts (50 ug, 12.4 ug and 2.5 ug) of ANTI-FLAG® M2 Monoclonal Antibody (Product Code: F3165) were spiked into COS-7 lysates (107 cells in 1 ml RIPA buffer). The mouse IgG was recovered using standard protein A agarose (White, lanes 1-3) or EZview™Red Protein A Affinity Gel (Red, lanes 4-6) in a mock IP experiment. After washing, the bound proteins were eluted and subjected to SDS-PAGE. The gels was stained with colloidal blue stain (EZBlue™ Gel Staining Reagent, Product Code: G1041).
Ordering Information:
Product # Product Name
E6654 EZview Red Anti-c-Myc
F2426 EZview Red Anti-FLAG® M2
E6406 New EZview Red Glutathione
E6779 EZview Red Anti-HA
E3528 EZview Red HIS-Select® HC
P6486 EZview Red Protein A
E3403 EZview Red Protein G
E5529 EZview Red Streptavidin
back to top
back to Purification & Detection