|
Combine IMAC Versatility with the Sensitivity of FLAG® Detection
The MAT-Tag™ System (metal affinity tag) utilizes a small, novel, seven amino acid (HNHRHKH) sequence created for IMAC purification of recombinant MAT fusion proteins and the high sensitivity detection of the antibody based FLAG® System.
- Dual tagged fusion proteins
- N- or C- terminal tagging configurations
- Compatible to Nickel and Cobalt affinity resins, such as HIS-Select
- Superior antibody detection
Data
Technical Posters
MAT Bacterial Expression Vectors
tac Promoter System
T7 Promoter System
T7 Promoter System with Enterokinase Recognition Site
MAT Mammalian Expression Vectors
Transient Expression
MAT Baculovirus Transfer Vectors
MAT Monoclonal Antibody
FLAG Antibody
Data
 |
 |
| Figure 1. Full-length FLAG-BAP-MAT fusion protein is detected on Western blots
by monoclonal antbodies to the FLAG and MAT tags. Lysates from uninduced (lanes 2 and 5) and induced (lanes 3 and 6)
E. coli cultures and ColorBurst Markers (lanes 1 and 4) were seperated by SDS-PAGE and blotted to nitrocellulose. The
resulting blot was immunostained using either ANTI-FLAG® M2-HRP conjugate (lanes 1-3) or Anti-MAT monoclonal antibody
(0.5 µg/ml) followed by Rabbit-Anti-Mouse IgG-HRP conjugate (lanes 4-6). The blots were developed and visualized with
TMB Substrate. |
Figure 2. MAT-tagged fusion protein expressed in E. coli can be affinity purified by IMAC. The coding region for GrpE was cloned into an E. coli expression plasimd, pFLAG-MAC, with an N-terminal FLAG tag and a C-terminal MAT tag. The fusion protein was expressed in E. coli strain BL21. A lysate (L, lane 2) was made in CelLytic B and separated by centrifugation into a pellet (P, lane 3) and a supernatant (S, lane 4) fraction. The supernatant was applied to a 1.0 mL HIS-Select HF Nickel Affinity Gel column and the flow through (F, lane 5), wash (1-3, lanes 6-8) and elution (1-4, lanes 9-12) fractions were collected. Samples of the fractions and ColorBurst Markers (lane 1) were seperated by SDS-PAGE and visualized by EZBlue staining. |
back to top
MAT Bacterial Expression Vectors
tac Promoter System
Vectors utilizing the strong tac promoter (a hybrid of the E. coli trp and lac promoters) offer protein expression levels in excess of 10 mg/L of culture when using IPTG as a de-repressor. These vectors can be used to express protein in any established E. coli host.
|
pTAC-MAT-1
Cytoplasmic expression of N-terminal MAT fusion proteins under the tac promoter. Supplied with a pTAC-MAT-2+BAP Control Vector.
| Product # |
Product Name |
Size |
| E5530 |
pTAC-MAT-1 Expression Vector |
10 µg |
|
 |
|
pTAC-MAT-2
Cytoplasmic expression of C-terminal MAT fusion proteins under the tac promoter. Supplied with a pTAC-MAT-2+BAP Control Vector.
| Product # |
Product Name |
Size |
| E5405 |
pTAC-MAT-2 Expression Vector |
10 µg |
|
 |
back to top
MAT Bacterial Expression Vectors
T7 Promoter System
The pT7-MAT vectors offer the very strong T7/lac promoter. These expression vectors produce even higher yields of recombinant proteins than the tac promoter system. However, the T7 promoter is known for background (“leaky”) expression, which can be a drawback when recombinant proteins are toxic to the host cell. Therefore, Sigma’s vectors contain the lac operator (lacO) sequences immediately downstream from the promoter to reduce leaky expression. Unlike the tac promoter system, pT7 vectors must be expressed in hosts containing a source of the T7 polymerase such as (DE3) lysogenic strains.
 |
pT7-MAT-1
Cytoplasmic expression of N-terminal MAT fusion proteins under the T7llac promoter. Supplied with a pT7-MAT-2+BAP Control Vector.
| Product # |
Product Name |
Size |
| E5780 |
pT7-MAT-1 Expression Vector |
10 µg |
|
 |
pT7-MAT-2
Cytoplasmic expression of C-terminal MAT fusion proteins under the T7llac promoter. Supplied with a pT7-MAT-2+BAP Control Vector.
| Product # |
Product Name |
Size |
| E5655 |
pT7-MAT-2 Expression Vector |
10 µg |
|
 |
pT7-FLAG-MAT-1
Cytoplasmic expression of N-terminal Met-FLAG, C-terminal MAT dual tagged fusion proteins under the T7llac promoter. Supplied with a pT7-FLAG-MAT-1+BAP Control Vector.
| Product # |
Product Name |
Size |
| E5280 |
pT7-FLAG-MAT-1 Expression Vector |
10 µg |
|
 |
pT7-MAT-FLAG-2
Cytoplasmic expression of N-terminal MAT, C-terminal FLAG dual tagged fusion proteins under the T7llac promoter. Supplied with a pT7-FLAG-MAT-1+BAP Control Vector.
| Product # |
Product Name |
Size |
| E4905 |
pT7-MAT-FLAG-2 Expression Vector |
10 µg |
|
back to top
MAT Bacterial Expression Vectors
T7 Promoter System with Enterokinase Recognition Site
The recognition sequence for enterokinase, Asp-Asp-Asp-Asp-Lys, is found at the C-terminal end of the FLAG® epitope tag. Removal of the FLAG is possible in all fusion proteins containing an N-terminal FLAG sequence. Dual tag fusion proteins may also be cleaved with enterokinase for removal of one or more tags, depending on the position of FLAG in the protein sequence.
|
pT7-MAT-FLAG-1
Cytoplasmic expression of N-terminal MAT-FLAG dual tagged fusion proteins under the T7llac promoter. Supplied with a pT7-FLAG-MAT-1+BAP Control Vector.
| Product # |
Product Name |
Size |
| E5155 |
pT7-MAT-FLAG-1 Expression Vector |
10 µg |
|
 |
|
pT7-FLAG-MAT-2
Cytoplasmic expression of C-terminal FLAG-MAT dual tagged fusion proteins under the T7llac promoter. Supplied with a pT7-FLAG-MAT-1+BAP Control Vector.
| Product # |
Product Name |
Size |
| E5030 |
pT7-FLAG-MAT-2 Expression Vector |
10 µg |
|
 |
back to top
MAT Mammalian Expression Vectors
Transient Expression
CMV vectors contain the pMB1 (derivative of pBR322) origin for replication in bacterial cells, the β-lactamase gene for ampicillin resistance selection in bacteria, hGH, polyA, and the f1 origin.
|
pCMV-FLAG-MAT-1
Transient cytoplasmic expression of N-terminal MeT-FLAG, C-terminal MAT dual tagged fusion proteins under the CMV promoter. Supplied with pCMV-FLAG-MAT-1 MAPK1 Control Vector.
| Product # |
Product Name |
Size |
| C5864 |
pCMV-FLAG-MAT-1 Expression Vector |
20 µg |
|
 |
|
pCMV-MAT-FLAG-1
Transient cytoplasmic expression of N-terminal MaT-FLAG dual tagged fusion proteins under the CMV promoter.
Supplied with pCMV-FLAG-MAT-1 MAPK1 Control Vector.
| Product # |
Product Name |
Size |
| C5989 |
pCMV-MAT-FLAG-1 Expression Vector |
20 µg |
|
 |
|
pCMV-FLAG-MAT-2
Transient cytoplasmic expression of C-terminal FLAG-MAT dual tagged fusion proteins under the CMV promoter.
Supplied with pCMV-FLAG-MAT-1 MAPK1 Control Vector.
| Product # |
Product Name |
Size |
| C6114 |
pCMV-FLAG-MAT-2 Expression Vector |
20 µg |
|
 |
back to top
MAT Baculovirus Transfer Vectors
pPolh-MAT are baculovirus transfer vectors used for producing Metal Affinity Tag (MAT-Tag) fusion proteins in insect cells. The pPolh-MAT vectors contain the strong viral polyhedrin (polh) promoter for high-level expression of target genes during the very late phase of infection. The vectors also contain a high copy of bacterial origin of replication and an ampicillin resistance gene (ampr) for easy propagation in E. coli host cells.
|
pPolh-MAT-1 Features
| Feature |
Map Position |
| AcNPV Sequence (ORF 603) |
1-1146 |
| Recommended 5' primer sequence binding site |
1079-1100 |
| polh Promoter |
1076-1145 |
| MAT |
1167-1187 |
| MCS |
1188-1246 |
| Recommended 3' primer sequence binding site |
1300-1320 |
| M13 origin |
2576-3229 |
| poly A |
1599-1604 |
| AcNPV Sequence (ORF 1629) |
1286-2629 |
| β-lactamase (amp') |
3616-4473 |
| pUC ori |
4624-5267 |
|
pPolh-MAT-2 Features
| Feature |
Map Position |
| AcNPV Sequence (ORF 603) |
1-1146 |
| Recommended 5' primer sequence binding site |
1079-1100 |
| polh Promoter |
1076-1145 |
| MCS |
1148-1211 |
| MAT |
1212-1232 |
| Recommended 3' primer sequence binding site |
1295-1315 |
| M13 origin |
2571-3224 |
| poly A |
1594-1599 |
| AcNPV Sequence (ORF 1629) |
1281-2624 |
| β-lactamase (amp') |
3611-4468 |
| pUC ori |
4619-5262 |
|
| Product # |
Product Name |
Size |
Application |
| T6699 |
pPolh-MAT-1 Transfer Vector |
20 µg |
Expression of N-terminal MAT fusion proteins in insect cells |
| T6574 |
pPolh-MAT-2 Transfer Vector |
20 µg |
Expression of C-terminal MAT fusion proteins in insect cells |
MAT Antibody
| Product # |
Product Name |
Size |
Characteristics |
Application |
| M6693 |
Monoclonal Anti-MAT antibody produced in mouse |
200 µg |
Specificity: N-terminal and C-terminal MAT fusion proteins |
- Immunoprecipitation
- Immunocytochemistry
- Western blotting
- EIA
Working Dilution
1 µg/ml by indirect Western blotting (chemiluminescent) |
FLAG Antibody
| Product # |
Product Name |
Package Size |
Characteristics |
Applications |
| F3165 |
ANTI-FLAG® M2 Monoclonal Antibody, Purified IgG |
200 µg 1 mg 5 mg |
Specificity: N-terminal, Met-N-terminal, Carboxy-terminal, or internal. Binding is not Ca2+-dependent.
Form: Solution in 10 mM sodium phosphate, 150 mM NaCl, pH 7.4, containing 0.02% sodium azide. |
Immunoprecipitation
Immunocytochemistry
Western blotting
EIA Working Dilution:
10 µg/ml by indirect Western blotting (chemiluminescent) |
For more information on our FLAG product offering, visit the FLAG System: Recombinant Protein Purification, Detection and Expression page.
back to top
back to Purification & Detection
|