Proteomics and Protein Expr.
Protein Expression
Cloning and Expression
Cell Lysis
Protocols
Purification and Detection
Protein Analysis
Stable Isotopes
Stem Cell Biology
Your Favorite Gene - Search
Life Science Innovations
PathFinder
 Product List for "Screening Kinase Phosphorylation Motifs Using Peptide Libraries" Protocol
Sigma-Aldrich Research Biotechnology
The following Cold Spring Harbor Laboratory Press protocols are brought to you by BioSupplyNet in a partnership with Sigma-Aldrich. A list of products is available to use in each protocol.
View the protocol on BioSupplyNet.com.
Selected Protocol from "Protein-Protein Interactions", 2nd Edition, E. Golemis, P. Adam.
Screening Kinase Phosphorylation Motifs Using Peptide Libraries
Isaac A. Manke and Michael B. Yaffe
Center for Cancer Research, Massachusetts Institute of Technology, Cambridge, MA
ABSTRACT
Determining which protein kinases are responsible for phosphorylating which protein substrates is critical for mapping signal transduction cascades that regulate cell growth, division, differentiation, and death. Oriented peptide library screening, combined with bioinformatics-based searches of protein sequence databases, offers a strategy for identifying substrates of particular protein kinases.
Buy this book, Z719145
Products Available for this Protocol
Protocol Material Description Product #  Product Name Add to Cart
Acetic acid, 0.1 m in water 45754 Acetic acid solution
Acetic acid, 30% in water 45754 Acetic acid solution
Acetone 00570 Acetone
Acetonitrile 92670 2,4,4-Trimethyl-1-pentene
Ammonium acetate, 0.1% (pH 11.5), freshly prepared and stored at 4°C A1542 Ammonium acetate
Ammonium acetate, 0.1% (pH 8.0), freshly prepared and stored at 4°C A1542 Ammonium acetate
Ammonium acetate, 0.1% (pH 9.5), freshly prepared and stored at 4°C A1542 Ammonium acetate
ATP, 10 mm (pH 7.0) (store at –20°C) A6559 Adenosine 5'-triphosphate disodium salt solution
DEAE Sephacel (Sigma I6505) I6505 Diethylaminoethyl–Sephacel™
Degenerate Ser- or Thr-containing peptide libraries Custom Peptides Custom Peptides  
EDTA, 0.1 m (pH 8.0) E7889 Ethylenediaminetetraacetic acid disodium salt solution
FeCl3, 100 mm in 0.1 m acetic acid (prepare fresh) 451649 Iron(III) chloride
Immobilized iminodiacetic acid agarose (IDA agarose) with 1,4 butanediol diglycidyl ether spacer I4758 Iminodiacetic acid Agarose
Methanol, dry M3641 Methanol
Phosphoric acid, 85% 452289 Phosphoric acid
Solution 1: 50 mm 2-(N-morpholino)ethanesulfonic acid (MES) (pH 5.5) containing 1 m NaCl M3671 MES
Standard reagents for peptide synthesis and protein sequencing (if this is being done in-house) Standard Reagents Standard reagents for peptide synthesis and protein sequencing  
Thionyl chloride 447285 Thionyl chloride
back to top
View all available Cold Spring Harbor Laboratory Press Protocols
Product Association Disclaimer: The Sigma-Aldrich products listed for this specific protocol were selected either to match or to supplement the products listed within the actual protocol. The products/reagents from Sigma-Aldrich have been qualified for usage, but may not have been validated for this specific application. Please refer to the detailed product description on the usage of specific products of interest.