Proteomics and Protein Expr.
Protein Expression
Protein Analysis
Stable Isotopes
Stem Cell Biology
Your Favorite Gene - Search
Life Science Innovations
PathFinder
 Panorama Functional Protein Arrays: Studying Cancer, Protein-Protein Interaction and Cell Signaling
Arrays
Panorama™ Functional Protein Arrays utilize a proprietary tagging technology based on the presence and biotinylation of a BCCP (biotin-carboxy carrier protein) tag. Panorama Functional Protein Arrays can be used to identify cellular response to external signals (cell signaling), researching protein involvement in cancer development and identifying protein-protein interaction and phosphorylation events.