|
Panorama® Antibody (Ab) Microarray – Gene Regulation I Kit is designed for studying protein expression in cell or tissue extracts. The antibody array contains 112 different antibodies each spotted in duplicate on a nitrocellulose-coated slide. These antibodies represent families of proteins found in the nucleus that are involved in chromatin remodeling and regulation of gene expression and include transcription factors, histone modifying enzymes, and antibodies that specifically recognize histone modification.
- Accurate and robust protein expression profiling of your sample in less than half a day
- Establish protein expression profile comparisons of two samples (test vs. reference samples), simply label Cy3 or Cy5 dyes to sample and apply simultaneously to array
- Proprietary slide treatment minimizes background staining
- Suitable for use with Cy3/Cy5 –based detection chemistries
Array Map (91 Kb PDF)
Slide Analysis Software (3 Kb GAL file)
Antibody List (95 Kb PDF)
Frequently Asked Questions (1.59 Mb PDF)
View Recent Literature
Order Information
| Product Code |
Product Description |
Technical Bulletin |
|
GRAA2 |
Panorama® Antibody Microarray – Gene Regulation I Kit |
|
back to top
back to Antibody Arrays
back to Proteomics
back to Enzyme Explorer
|