Accurately identify your target's interaction against 130 fully functional cancer proteins with the Panorama Cancer Protein Array! Based on a proprietary tagging technology that ensures only functional proteins are immobilized, the cancer protein array is an essential research tool for unveiling details of protein involvement in cancer development.
|
- Accurate representation of 130 cancer proteins.
- Proteins are expressed in insect cells and analyzed by Western blot to verify generation of
full-length proteins.
- Compatible with probing by DNA, proteins, kinases & small molecules and with Cy3/Cy5 –based detection chemistries.
Functional Array Technology
Cancer Protein Content (192 Kb Excel file)
Slide Analysis Software (6.71 Kb GAL file)
Slide Analysis Template (2.01 Mb Excel file)
Order Information
| Product Code |
Product Description |
Technical Bulletin |
|
HPFM2 |
Panorama® Human Cancer v1 Protein Array- |
|
 |
Kit Contents:
- Panorama Human Cancer v1 Protein Array - 2 ea
- Calmodulin - Cy5
- Monoclonal Anti-c-Myc-Cy3 Conjugate
- Cover slips
- quadiPERM Vessel
- 50 mL Conicol - 2 ea
- Pap Jar - 4 ea
- Assay Buffer
- Calmodulin Buffer
- BSA
- 1M DTT
- Cancer v1 Array Diskette (GAL file and analysis spreadsheet)
|
This product is sold under an exclusive worldwide license and a joint collaborative agreement between Sigma-Aldrich Corp. and Procognia Limited.
Online survey offer is valid only for qualified existing or new Sigma-Aldrich customers.
back to top
back to Proteomics
back to Cancer Research
back to Enzyme Explorer
|