|
Panorama® Antibody (Ab) Microarray – MAPK & PKC Pathways Kit is designed for studying protein expression in cell or tissue extracts. The antibody array contains 84 different antibodies each spotted in duplicate on a nitrocellulose-coated slide. The antibodies are specific for kinases and phosphatases of the MAPK and PKC pathways.
- Accurate and robust protein expression profiling of your sample in less than half a day
- Establish protein expression profile comparisons of two samples (test vs. reference samples), simply label Cy3 or Cy5 dyes to sample and apply simultaneously to array
- Proprietary slide treatment minimizes background staining
- Suitable for use with Cy3/Cy5 –based detection chemistries
Array Map (87 kb PDF)
Slide Analysis Software (3 kb GAL file)
Antibody List (94 kb PDF)
Worksheet for Data Analysis and Calculations (17 kb PDF)
Frequently Asked Questions (1.59 Mb PDF)
View Recent Literature
Order Information
| Product Code |
Product Description |
Technical Bulletin |
|
MPAA3 |
Panorama® Antibody Microarray – MAPK and PKC Pathways |
|
back to top
back to Antibody Arrays
back to Proteomics
back to Enzyme Explorer
|