|
Panorama® Antibody Microarray – p53 Pathways Kit is designed for studying protein expression in cell or tissue extracts. The array contains 112 different antibodies each spotted in duplicate on a nitrocellulose-coated slide. The immobilized antibodies represent various p53 pathways and are also available independently.
- Accurate and robust protein expression profiling of your sample in less than half a day
- Qualitative characterization of protein expression against 112 different antibodies
- Establish comparative protein expression profiles between two samples
- Proprietary slide treatment minimizes background staining
- Suitable for use with Cy3/Cy5 – based detection chemistries
Antibody List (64.5 kb PDF)
Slide Analysis Software (3 Kb GAL file)
Array Map (7.75 kb PDF)
Frequently Asked Questions (1.59 Mb PDF)
View Recent Literature
Order Information
| Product Code |
Product Description |
Technical Bulletin |
|
PPAA4 |
Panorama® Antibody Microarray – p53 Pathways |
|
 |
Kit Contents:
Panorama® Antibody Slides – p53 Pathways (2 Each)
quadriPERM Cell Culture Vessel (2 Each)
Extraction/Labeling Buffer
Protease Inhibitor Cocktail
Phosphatase Inhibitor Cocktail II
Benzonase®, Ultrapure
Array Incubation Buffer
Phosphate Buffered Saline, pH 7.4, with Tween® 20 (Washing Buffer)
SigmaSpin Post-Reaction Clean-Up Columns
Collection Tubes, Polypropylene, 2 ml
Panorama® Antibody List – p53 Pathways |
QuadriPERM is a trademark of WC Heraeus GmbH, Ltd.
Tween is a registered trademark of the ICI Group.
Benzonase is a trademark of Merck KGaA.
Cy3 is a trademark of GE Healthcare.
Panorama is a registered trademark of Sigma-Aldrich.
Antibody Arrays
Proteomics
Cell Signaling
Antibody Explorer
|