|
The GlycoProfile Labeling Kits are designed for efficient labeling of
N-linked, O-linked and glycosylphosphatidylinositol (GPI) anchored glycans using your
choice of 2-aminobenzamide (2-AB) or 2-aminobenzoic acid (anthranilic acid; 2-AA)
1,2. These small fluorophores increase the spectral absorption of glycans,
improving detection in HPAE (high-performance anion exchange chromatography),3,4,5,6
HPLC4,5,6,7, and ESI-MS (electrospray mass spectroscopy) methods. Binding is
robust and the resulting dye-glycan conjugates are stable, with no degradation during
post-labeling analysis. 2-AB is recommended for downstream analysis by HPAE, HPLC and
ESI-MS, although it is slightly less sensitive than 2-AA. 2-AA is recommended for
SDS-PAGE, and is also suitable for HPAE, HPLC and ESI-MS.
Labeling of carbohydrates with 2-AB
Features and Benefits
- Each kit is complete, containing 2-AA or 2-AB dye, reductant and solvents.
- Sufficient for the preparation of up to 36 samples of mixed glycans containing
from 100 picomoles to 50 nanomoles of purified glycans. With a single pure glycan, as
little as 5 picomoles may be labeled and detected in subsequent HPLC analysis.
Note: Labeling of glycans with 2-AB is covered under US Patent No
5,747,347 and its foreign equivalents.
GlycoProfile Glycan Clean-Up Cartridges are recommended for use
with the GlycoProfile Labeling kits for post-labeling cleanup. Glycans adsorb to the
membrane while excess dye, mono- and disaccharides and other hydrophobic contaminants
pass through. Purified labeled glycans are eluted with water and are ready for
subsequent enzymatic digestion or analysis.
Ordering Information
| Product Code |
Product Name |
Unit |
Technical Bulletin |
| PP0530 |
GlycoProfile 2-AA Labeling Kit |
1 kit |
|
| PP0520 |
GlycoProfile 2-AB Labeling Kit |
1 kit |
|
| G8169 |
GlycoProfile Glycan Clean-up Cartridges |
3 each
6 each
12 each |
|
References:
- Bigge, J.C., et al., Non-selective and efficient fluorescent labeling of
glycans using 2-aminobenzamide and anthranilic acid, Anal. Biochem., 230,
229-238 (1995).
- Hardy, M.R., Glycan labeling with the fluorophores 2-aminobenzamide and
anthranilic acid, Techniques in Glycobiology, Dekker, Inc. NY (1997).
- Delaney, J., and Vouros, P., Liquid chromatography ion trap mass spectrometric
analysis of oligosaccharides using permethylated derivatives, Rapid Commun. Mass
Spectrom., 15(5), 325-334 (2001).
- Anumula, K.R., and Dhume, S.T., High resolution and high sensitivity methods for
oligosaccharide mapping and characterization by normal phase high performance liquid
chromatography following derivitization with highly fluorescent anthranilic acid,
Glycobiology, 8, 685-694 (1998).
- Townsend, R., et al., Multimode high-performance liquid chromatography of
fluorescently labeled oligosaccharides from glycoproteins, Anal. Biochem., 239,
200-207 (1996).
- Royle, L., et al., An Analytical and Structural Database Provides a
Strategy for Sequencing O-Glycans from Microgram Quantities of Glycoproteins, Anal.
Biochem., 304, 70-90 (2002).
- Guile, G.R., et al., A rapid and high-resolution high-performance liquid
chromatographic method for separating glycans mixtures and analyzing oligosaccharide
profiles, Anal. Biochem., 240, 210-226 (1996).
- Rudd, P.M., et al., Glycoproteins: rapid sequencing technology for
N-linked and GPI Anchor glycans, Biotechnol. Genet. Eng. Rev., 16,
1-21(1999).
|