Proteomics and Protein Expr.
Protein Expression
Protein Analysis
Stable Isotopes
Stem Cell Biology
Your Favorite Gene - Search
Life Science Innovations
PathFinder
 ProteoPrep® 20 Plasma Immunodepletion LC Column ProteoPrep 20  - XML/RSS Feed
Proteomics
Red cube and magnifying glass   Discover Protein Biomarkers Faster
ProteoPrep 20 Plasma Immunodepletion LC Column
ProteoPrep® 20 Plasma Immunodepletion LC Column (PROT20LC)
By removing 20 highly abundant proteins (representing 97% of the proteome) at 98% efficiency, less fractionation of the true plasma proteome is required; focusing on your protein biomarkers becomes faster and simpler!
  • Reduce sample complexity and increase efficiency
  • Reveal proteins previously masked
  • Enrich proteins of interest up to 20-fold
Designed for use with LC instruments, the ProteoPrep® 20 LC column accommodates a sample load of 100 µL plasma or serum and can be used up to 100 times.
Read the press release:
Biomarker Discovery Simplified with Innovative Technology from Sigma-Aldrich
ProteoPrep 20 Plasma Immunodepletion Kit Components
  • 1 Depletion Column
  • 10X Equilibration Buffer
  • 10X Elution Buffer
  • Kathon (for long-term storage)
  • Collection Tubes
  • Spin Filters
ProteoPrep 20 Technology Button
Resources Button
Related Products Button
Ordering Information
Product # Product Name Package Size
PROT20LC ProteoPrep® 20 Plasma Immunoaffinity LC Column 1 kit
ProteoPrep® 20 Plasma Immunodepletion Technology
The ProteoPrep® 20 technology offers the highest depletion of highly abundant plasma proteins available today and specifically removes 20 highly abundant proteins (representing 97% of the proteome) to reveal a new layer of the plasma proteome.
The ProteoPrep® 20 Technology depletes the following proteins:
Albumin
IgGs
Transferrin
Fibrinogen
IgAs
α-2-Antitrypsin
IgMs
α-1-Antitrypsin
Complement C3
Haptoglobulin
Apolipoprotein A1
Apolipoprotein A2
Apolipoprotein B
Acid-1-Glycoprotein
Ceruloplasmin
Complement C4
Complement C1q
IgDs
Prealbumin
Plasminogen
Twenty-two Unique Proteins Revealed with ProteoPrep® 20
Twenty-two Uniques Proteins Revealed with ProteoPrep 20
The ProteoPrep® 20 Plasma Immunodepletion Kit specifically removes 20 highly abundant proteins (representing 97% of the proteome) at 99% efficiency, revealing a new layer of highly abundant proteins. Initially, the gel image above appears to be similar to an undepleted sample, but identifications via mass spectrometry indicate that the visible proteins are not members of the list of twenty high abundance proteins specifically removed. They are, indeed, unique to a sample depleted with the ProteoPrep® 20 technology.
Spot No. Protein Identification
1
2
3
4
5
6
7
8
9
10
11
12
13
14
15
16
17
18
19
20
21
22
Alpha-1-B-glycoprotein
Hemopexin (Beta-1B-glycoprotein)
Alpha-2-HS-glycoprotein
(Fetuin A)
Vitamin D Binding Protein
Pigment Epithelium-derived factor
Carboxypeptidase N
(Kininase 1)
Complement Factor H fragment
Apolipoprotein E
Ficolin 3 (Hakata antigen)
PK-120 (Plasma Kallikrein-sensitive Glycoprotein)
Apolipoprotein A-IV
Glutathione Peroxidase 3
Tetranectin (Plasminogen Binding Protein)
Serum amyloid A (SAA)
Kininogen
Complement Factor H-related protein 2
Inter-alpha-trypsin inhibitor H1
Vitronectin
Geltain-Binding Protein
Antithrombin III
Cluterin (Apolipoprotein J)
Inter-alpha-trypsin inhibitor H4
Back to Top
Resources
Typical Depletion Chromatogram
Typical Depletion Chromatogram
ProteoPrep 20 Plasma Immunodepletion LC Column User Guide PDF
ProteoPrep® 20 Plasma Immunodepletion LC Column User Guide (277 Kb PDF)
Non-Specifically Bound Proteins and Peptides Poster
Examination of Non-Specifically Bound Proteins and Peptides Following Depletion of Twenty High Abundance Proteins from Human Plasma (211 Kb)
Presented by Chris Melm at ABRF, 2007
Lower Abundance Proteome Poster
Absolute Quantification of the Lower Abundance Proteome Through Immunoaffinity Depletion of the Twenty Most Abundant Proteins in Human Serum (466 Kb PDF)
Presented by Jeff Porter at US HUPO, 2007
Comparison of Precipitation Methods Poster
Comparison of Precipitation Methods Following Depletion of Twenty High Abundance Proteins from Human Plasma (204 Kb PDF)
Presented by Mark Schuchard at PEP-talk, 2006
Back to Top
Related Products
ProteoPrep® 20 Plasma Immunodepletion Kit
ProteoPrep® 20 spin columns specifically remove 20 of the most abundant proteins from human plasma or serum to reveal previously masked protein biomarkers. Additionally, this facilitates a 20-fold increase in the relative amount of revealed proteins that can be subsequently studied and analyzed. Designed for use with bench top centrifuges, ProteoPrep® 20 spin columns can accommodate a sample load of 8 µL plasma or serum and can be used up to 100 times each. The ProteoPrep® 20 Plasma Immunodepletion Kit contains 3 spin columns. The ProteoPrep® 20 Sample Kit contains 1 spin column.
ProteoPrep 20 Plasma Immunodepletion Kit
Components
  • Depletion Spin Column(s)
  • 10X Equilibration Buffer
  • 10X Elution Buffer
  • Kathon (for long-term storage)
  • Collection Tubes
  • Spin Filters
  • Syringes
  • Luer Lock Caps
Product # Product Name Package Size
PROT20 ProteoPrep® 20 Plasma Immunodepletion Kit 1 kit (3 columns)
PROT20S ProteoPrep® 20 Plasma Immunodepletion Sample Kit 1 kit (1 column)
ProteoPrep® Blue Albumin & IgG Depletion Kit
ProteoPrep Blue Albumin Depletion Kit is designed to specifically remove albumin and IgG from 25 samples of human serum (25 µl to 100 µl) in preparation for two-dimensional electrophoresis (2DE). Albumin-depleted serum samples generated by this kit are in urea rather than salt-based buffers with little dilution, eliminating the need to precipitate the sample prior to performing 2DE.
ProteoPrep 20 Blue Albumin & IgG Depletion Kit
Components
  • ProteoPrep® Blue Albumin and IgG Depletion Medium—a 10-ml suspension containing 60% packed medium
  • ProteoPrep® Blue Equilibration buffer—1 bottle reconstitutes to 50 ml, pH 7.8
  • Protein Extraction Reagent Type 4—1 bottle reconstitutes to 23 ml, pH 10.4
  • 25 Spin columns
  • 75 Collection Tubes
Product # Product Name Package Size
PROTBA ProteoPrep® Blue Albumin & IgG Depletion Kit 1 kit
Back to Top