Proteomics and Protein Expr.
Protein Expression
Protein Analysis
Protein Quantitation
Protein Staining/Detection
Protein Electrophoresis
Western Blotting
Mass Spectrometry
X-Ray Crystallography
Chromatography
Stable Isotopes
Stem Cell Biology
Your Favorite Gene - Search
Life Science Innovations
PathFinder
 ProteoSilver™ and ProteoSilver Plus
Protein Electrophoresis
Highly sensitive ProteoSilver and ProteoSilver Plus are ideal products for any proteomics scientist. Both kits contain prepared solutions of silver staining reagents along with detailed instructions to achieve optimal results. With a detection limit of 0.1 ng/mm 2 of protein (BSA) and an extremely low background, ProteoSilver leads to superior detection of low abundance proteins. ProteoSilver Plus is MALDI-MS compatible and does not contain glutaraldehyde, which crosslinks lysine residues, resulting in inaccurate spectra. This kit contains two additional reagents for destaining an excised protein spot, for those wishing to perform further characterization through tryptic digest and MALDI-MS analysis. Both kits contain sufficient reagents for staining 25 mini-gels.
Features & Benefits
  • Pre-mixed and pre-weighed solutions reduce time and cost of purchasing and preparing individual components
  • High sensitivity and low background ensure very low abundance proteins can be detected and resolved from other proteins
  • MALDI-compatible. Protein spots of interest can be further characterized by mass spectrometry using ProteoSilver Plus
ProteoSilver and ProteoSilver Plus Staining Kits Image
Flow Diagram for ProteoSilver Plus Destaining
Flow Diagram for ProteoSilver™ and ProteoSilver Plus Staining
Protein Stains Selection Chart
Product #  Product Name Size
PROT-SIL1 ProteoSilver 1 kit
PROTSIL2 ProteoSilver Plus 1 kit
back to top
back to Protein Electrophoresis