Proteomics and Protein Expr.
Protein Expression
Protein Analysis
Protein Quantitation
Protein Staining/Detection
Protein Electrophoresis
Western Blotting
Mass Spectrometry
X-Ray Crystallography
Chromatography
Stable Isotopes
Stem Cell Biology
Your Favorite Gene - Search
Life Science Innovations
PathFinder
 QuantiPro™ High Sensitivity Protein Assay Kit
Protein Quantitation
Why settle for common methods?
Achieve superior performance with QuantiPro™ Protein determination is one of the most common types of analysis performed in biochemical research. That doesn't mean you should settle for common methods. A new generation of protein assay kits from Sigma-Aldrich has arrived. The QuantiPro bicinchoninic acid (BCA) Assay Kit (QP-BCA) is the perfect match for high-throughput screening and multiwell assays. Because it's more sensitive than other procedures, QuantiPro provides more accurate results, with less variability. You gain the edge with precise, accurate, reproducible results - every time.
It's all about dependability.
QuantiPro relies on the formation of a copper-protein complex under alkaline conditions. BCA forms a purple-blue complex, which provides the basis to monitor the amount of protein present. The color complex is stable, so you can step away from your research with the knowledge that accurate results await your return. In addition, BCA is the reliable, preferred method for quantitation in the presence of detergents.
Each QuantiPro BCA Kit Contains:
QuantiPro Buffer QA
QuantiPro BCA QB
Copper(II) sulfate, pentahydrate, 4% (w/v) solution
Protein Standard Solution
250 mL
250 mL
12 mL
10 ampules of 1.0 mL each
BCA Protein Compatibility Chart
Ordering Information
Product #  Product Name Size
QP-BCA QuantiPro BCA Assay Kit 1 kit
back to top
back to Protein Quantitation