|
Sigma offers a variety of prestained markers as well as a biotinylated marker for use in Western blotting analysis. Prestained markers contain proteins conjugated to brilliantly colored dyes, and Sigma's biotinylated marker contains biotin conjugated proteins easily detected with a Streptavidin Peroxidase conjugate and color development reagent.
| Product # |
Product Name |
MW Range (Da) |
|
SDS7B2 |
Prestained Molecular Weight Marker |
(27,000-189,000) |
|
C4861 |
ColorBurst Electrophoresis Marker
|
High (30,000-220,000) |
|
C1992 |
ColorBurst Electrophoresis Marker
|
(8,000-220,000) |
|
C3187 |
Color Marker |
Low Range (6,500-45,000) |
|
C6210 |
Color Marker |
Ultra-Low Range (1,060-26,600) |
|