Proteomics and Protein Expr.
Protein Expression
Cloning and Expression
Cell Lysis
Protocols
Purification and Detection
Protein Analysis
Stable Isotopes
Stem Cell Biology
Your Favorite Gene - Search
Life Science Innovations
PathFinder
 HIS-Select® Technology
Purification & Detection
HIS-Select Technology Header
Sigma's HIS-Select products have the ability to purify histidine-tagged proteins quickly and with high selectivity. This is due to HIS-Select's patented, non-charged, hydrophilic nickel chelate linkage. This non-charged linkage greatly diminishes non-specific binding. Most other systems for histidine-tagged protein purification utilize a charged chelate linkage. The tetradentate chelator used in the HIS-Select products results in greatly reduced nickel leaching and in high specificity for purification or affinity capture of your histidine-tagged protein. In addition, HIS-Select eliminates the need for costly and time-consuming secondary purification of your protein, allowing for One-step-purification. Read more...
  • High Selectivity for Higher Purity
  • Non-charged, hydrophilic linkage reduces non-specific binding
  • Pure tetradentate chelate for minimal metal leaching
  • One-step purification
HIS-tagged proteins
What's New!
Resins Convenient Formats Plates
  • Nickel Affinity Gel
  • High Flow Nickel Affinity Gel
  • HIS-Select® Cobalt Affinity Gel
  • IMAC-Select Affinity Gel
  • HIS-Select iLAP® 5 ml Column
  • Cartridge
  • Spin Column
  • EZview™ Red Nickel Affinity Gel
  • iLAP® Plates
  • High Capacity Nickel Coated Plates
  • High Sensitivity Nickel Coated Plates
  • HIS-Select® Filter Plates
Kits Supporting Products & Reagents  
  • M Purification Kit
  • Y Purification Kit
  • M Affinity Capture Kit
  • Wash & Elution Buffer Kit
  • Trypsin
  • Maldi Kits
  • CelLytics
  • ProteoQwest Kits
  • ProteoSilver
  • HIS-Select® Wash & Elution Buffers
Technical Publications & Literature
Recent R&D Posters:
Life Science Quarterly Application Note:
Automation Protocols:
Additional Literature:
Use the "Login" button on the tool bar (at the top of the page) to be able to view package sizes and prices when you click on the product numbers.
back to Purification & Detection
HIS-Select and products are covered by US Patent No 6,623,655 and licensed under U.S. Patent No. 4,569,794 and are sold for research use only. Commercial use of Sigma's HIS-Select, PHOS-Select, iLAP and IMAC products require additional licenses.