|
ProteoProfile Enzymatic In-gel N-Deglycosylation Kit, (PP0200) |
 |
Protein Phosphorylation |
 |
 |
| |
|
|
|
Most proteins in eurkaryotic cells undergo post-translational modification,
quite commonly glycosylation. Unfortunately, this type of modification leads
to problems in subsequent protein analysis procedures. Glycopeptides generally
do not readily ionize during MS analysis leading to insufficient spectral
data. Furthermore, proteolytic digestion of the native glycoprotein is often
incomplete due to steric hindrance from the bulky oligosaccharides attached
to the protein molecule. Hence removal of the carbohydrate groups from a
glycoprotein prior to protein identification is preferred. |
Sigma's ProteoProfile Enzymatic In-gel N-Deglycosylation Kit is optimized
to provide a convenient and reproducible method to N-deglycosylate and tryptically
digest protein samples from 1D or 2D polyacrylamide gel slices for subsequent
MS or HPLC analysis. The procedure is suitable for Coomassie Blue and Colloidal
Coomassie stained gels and may be used with gels silver stained and destained
using Sigma's Proteo Silver Plus kit (
PROT-SIL2). ProteoProfile Enzymatic In-gel N-Deglycosylation kit includes the enzymes and reagents necessary for N-linked deglycosylation and tryptic digestion. The samples can then be desalted and concentrated for analysis by MALDI-TOF or electrospray MS.
Kit Components
Destaining Solution
Proteomics Grade PNGase F
Proteomics Grade Trypsin
Trypsin Solubilization Reagent
Trypsin Reaction Buffer
Ribonuclease B (positive control)
Peptide Extraction Solution
Biotech Grade Acetonitrile |
|
Features & Benefits
- Provides all components for in-gel deglycosylation AND trypsinization of protein samples
Conveniently prepares deglycosylated protein samples for analysis by MS or HPLC
- Utilizes PNGase F for the enzymatic removal of N-linked glycans
Proteins remain intact, unlike the use of chemical deglycosylation which can degrade the protein
- Includes Proteomics Grade PNGase F and Trypsin
Highly purified enzymes possess no unwanted activities or additives to complicate analysis.
- PNGase F is supplied lyophilized from a low salt buffer
Allows reconstitution of
the enzyme to any concentration needed
- Detailed protocol
Easy to follow protocol simplifies the process
- Works in solution or with gel slices
Allows choice of methods
View Protocol Diagram
back to top
back to Phosphorylation