| Product # |
Product Name |
Function |
Format |
Samples Processed |
Input Sample Size |
Output Compatibility |
| P9740 |
PHOS-Select Iron Affinity Gel |
Agarose Fe+3-chelate phosphopeptide capture |
Bulk media |
5 |
Captures 4 µM phosphopeptide maximum per mL packed media |
LC, MALDI-MS, ESI-MS |
| PP0410 |
PhosphoProfile I Phosphopeptide Enrichment Kit |
Silica Ga+3-chelate phosphopeptide capture |
Prepacked spin-column |
24 |
Captures 25 nM phosphopeptide maximum per spin column |
LC, MALDI-MS, ESI-MS |
PHOS-Select Iron Affinity Gel, (P9740)
- High capacity (at least 4 micromoles of phosphopeptide per ml of resin)
- Precharged with iron (III) as a stable chelate
- Provides single-step phosphocompound purification
- Supplied as an approximate 50% suspension in 50% glycerol buffered at pH 5.0
- Supplied with technical bulletin providing detailed procedure for use
View Protocol Diagram
Data
PhosphoProfile I Phosphopeptide Enrichment Kit, (PP0410)
Mass spectrometry (Matrix Assisted Laser Desorption/ Ionization Time of Flight (MALDI-TOF) or Electrospray Ionization (ESI)) of phosphopeptides from tryptic protein digests is a powerful tool for charactering and identification of phosphorylation sites. A combination of low intrinsic abundance, inefficient ionization, and/or signal suppression of most phosphopeptides may limit or even prevent detection, unless the phosphopeptide content is significantly enriched prior to analysis. This kit conveniently includes all materials needed to enrich for phosphopeptides from digested samples in a robust and unbiased manner (see binding comparison). The phosphopeptide capture matrix is a novel Ga+3 chelate silica based on a proprietary nitriloacetic acid (NTA) analog. The silica beads are approximately 20 microns in diameter with a pore size of 1,000 Angstroms. The matrix is packed into spin columns for easy, microscale affinity capture of phosphopeptides (see workflow).
The complete solution kit – 24 samples up to 25 nmoles phosphophopeptide each
- Mass spec compatible – save time, reduce sample handling and potential loss
- Proteomics Grade Trypsin - for clean and complete digests
- PHOS-Select Gallium Spin Columns – high capacity, novel Ga+3 silica media for fast, unbiased capture and recovery
- Controls – validate your process for confidence and reporting
- Buffers – enzyme reaction, binding, washing and elution optimized formulations for robust performance
- Consumable equipment included – matched equipment means no risk of sample loss, additional purchases or waste
Workflow highlighting the use of the spin columns for digestion and selective enrichment of phosphopeptides from sample.
Binding comparison: Selected portions of HPLC chromatograms demonstrating enrichment of phosphopeptides for IMAC technologies. Standard phosphopeptides representing the three most common sites of phosphorylation phosphoserine, phosphothreonine, and phosphotyrosine were used. The lyophilized solids were first dissolved in water and an approximately equimolar mixture of the peptides was formulated. Each phosphopeptide was added to the BSA digest at a weight ratio of ~1.7% to produce a total phosphopeptide content of ~5% by weight. Quantitation results are given below. Note that competitor A, B, and C technologies were biased in selecting Peptide 2, while Sigma’s technology bound and eluted the peptides in approximately the same ratio as applied to the column.
 |
| |
| Kit |
Recovery of phosphopeptide standards |
Specificity* |
| 1 |
2 |
3 |
Total |
| Sigma |
59% |
52% |
74% |
59% |
50% |
| Competitor A |
6% |
19% |
11% |
13% |
28% |
| Competitor B |
39% |
56% |
17% |
42% |
28% |
| Competitor C |
37% |
65% |
37% |
46% |
25% |
|
Additional Products for Phosphoprotein Analysis
| Immobilized Antibodies for the Isolation of Phosphopeptides |
|
| Product Name |
Product # |
| Monoclonal Anti-Phosphotyrosine-Agarose
from mouse, Purified immunoglobulin, PBS suspension, clone PT-66 |
A1806 |
| Monoclonal Anti-Phosphoserine-Agarose,
Purified immunoglobulin, PBS suspension, clone PSR-45 |
A8076 |
| Monoclonal Anti-Phosphothreonine-Agarose
from mouse, Purified immunoglobulin, PBS suspension |
A7951 |
| Controls and Elution Reagents |
back to top |
| Product Name |
Product # |
| Phosphotyrosine-BSA Phosphotyrosine
Antibody Inhibitor, 2 mg per ml, Buffered aqueous solution,
for blotting and ELISA |
P3967 |
| Phosphoserine-BSA, Phosphoserine Antibody
Inhibitor, 2 mg/ml, for blotting and ELISA |
P3717 |
| Phosphothreonine-BSA, Phosphothreonine
Antibody Inhibitor, 2 mg/ml, for blotting and ELISA |
P3842 |
| Sodium phenyl phosphate dibasic dihydrate,
95%, elutes phosphoproteins from antibody resins |
P7751 |
| Resins and Plates |
back to top |
| Product Name |
Product # |
| Avidin-agarose from egg white, Aqueous
glycerol suspension |
A9207 |
| Avidin (Monomeric)-Agarose from egg
white, Aqueous glycerol suspension |
A2036 |
| Streptavidin-Agarose from Streptomyces
avidinii, Buffered aqueous suspension |
S1638 |
| SigmaScreen ExtrAvidin Coated Plates |
E3027 |
| SigmaScreen Streptavidin 96-well High
Capacity Coated Plates |
S6940 |
| SigmaScreen Streptavidin 384-well High
Capacity Coated Plates |
S8815 |
| Anti-Mouse IgG (whole
molecule)-Agarose from goat, IgG fraction of antiserum, Saline
suspension |
A6531 |
| EZviewä
Red Protein A Affinity Gel |
P6486 |
| EZviewä
Red Protein G Affinity Gel |
E3403 |
| Protein L-Agarose from Peptostreptococcus
magnus, recombinant, expressed in Escherichia coli |
P3351 |
| Protein A coated HS 96-well plates,
flat-bottomed 96-well plates, clear polystyrene |
S1938 |
| Protein G coated HS 96-well plates,
flat-bottomed 96-well plates, clear polystyrene |
S2063 |
Phosphatase Inhibitor Cocktails
| Product Number |
P5726 |
P2850 |
| |
Tyrosine Protein Phosphatases and Acid and Alkaline Phosphatases |
Ser/Thr Protein Phosphatases and Alkaline Phosphatase L-Isozymes |
| (--)-p-Bromotetramisole |
|
 |
| Cantharidin |
|
 |
| Imidazole |
 |
|
| Microcystin LR |
|
 |
| Sodium molybdate |
 |
|
| Sodium orthovanadate |
 |
|
| Sodium tartrate |
 |
|
back to top
back to Phosphorylation
|