Life Science
Antibody Explorer
Automation
Cancer Research
Cell Culture
Cell Signaling
Custom Synthesis
Drug Discovery
Functional Genomics and RNAi
Product Lines
Related Products
Key Resources
Molecular Biology
Nutrition Research
Plant Biotechnology
Proteomics and Protein Expr.
Stable Isotopes
Life Science Quarterly
 MISSION™ shRNA Target Search Database
RNA Interference (RNAi)
Welcome to MISSION Control
Search the Clone Database
MISSION™ shRNA Target Search & Ordering Database
(RefSeq No., or Clone ID or TRC No.) : help
Gene Description or Symbol : help
Examples
RefSeq No.: NM 002227
Clone ID: NM_002227.x-898s1c1
TRC No.: TRCN0000003102
Gene Description: Janus kinase
Symbol: JAK1
   NCBI Database - Check NCBI for the latest RefSeq numbers associated with your target gene.
Locate shRNA sets for your specific gene of interest through the MISSION shRNA Target Search database. The database may be searched using RefSeq or Accession Numbers, TRC numbers, gene symbols, gene descriptions among other specific identifiers. Target sets found in the collection based on your search criteria will be returned on a Search Results Page and listed by RefSeq number. The results can be further refined and sorted or you may click on a target to obtain in depth information about the shRNA set. Please refer to the help files in the event you have trouble using the database or locating your target.
back to top
View Detailed Information about the shRNA Libraries
FAQs
shRNA Product Formats
Ordering Information
Contact Us
Send this page to a colleague.
shRNA Product Formats
The library is currently provided as frozen bacterial glycerol stocks harboring sequence-verified shRNA lentiviral plasmid vectors or as purified plasmid DNA. Each gene is represented by a target set that consists of an average of 3-5 individual constructs or clones targeting different regions of the gene sequence. Bacterial cultures may be amplified from the glycerol stocks for use in purification of the shRNA plasmid DNA. Alternatively, the purified plasmid format does not require these extra steps as the growth and DNA purification are completed for you. Target cell lines may be transfected with the purified plasmid for transient or stable gene silencing (puromycin selection). In addition, self-inactivating replication incompetent lentiviral particles can be produced in packaging cells (HEK 293T) by co-transfection with compatible packaging plasmids. Orders of 5 or less target sets (≤ 25 constructs) are provided in individual 1.4 ml TrakMates® tubes. Each TrakMates® tube contains a unique 2 dimensional barcode on the bottom of the tube that can be read using a corresponding reader. A printed value corresponding to the The RNAi Consortium (TRC) clone number is also provided on each tube. Orders of >5 clone sets (>25 constructs) are automatically provided in a 96-well plate with a one dimensional barcode label on the plate and are provided with a CD containing plate map positions.
MISSION shRNA Bacterial Glycerol Stock
MISSION shRNA Plasmid DNA
back to top
Gene Family Sets
Custom gene family sets are available at anytime. Please check with your local sales representative or e-mail us at RNAi@sial.com for more information.
Check back periodically for additional target sets, formats, gene family sets, and updates to the web search capabilities.
MISSION shRNA Sets
  • MISSION shRNA Human Kinases: Glycerol Set-1 - SH0111
Ordering Information
Clone sets of the MISSION shRNA library are ordered online through the MISSION shRNA Target Search database. To order online you will need to have a login profile for the website and request online order access in your profile. Upon completion of your profile, you will be able to add the products to your order cart and complete online transactions. Orders may be placed using Purchase Order numbers or credit cards. Pricing noted on the web is for 1-20 target sets. For pricing beyond 20 target sets, please contact your local sales representative, or e-mail us at RNAi@sial.com. Bacterial glycerol stocks and purified plasmid DNA are considered custom orders and are produced on-demand and shipped directly to end-users worldwide.
How to order target sets:
  1. Update or create your web login profile with order access privilege (one time required step that may take 24-48 hours to process).
  2. After logging in, search the database and add target sets to your cart.
  3. Check out and submit your order.
Note: If you are providing clone product numbers to a purchasing agent, please let them know the URL of the ordering database: www.sigma-aldrich.com/missionsearch.
back to top
Contact Us
For questions about the library, pricing and quotes or other concerns, please e-mail us at: RNAi@sial.com.
Back to RNAi
MISSION is a trademark of Sigma-Aldrich.
The MISSION shRNA Library of The RNAi Consortium is produced and distributed under license from the Massachusetts Institute of Technology.
Licensed under Carnegie Institution US Patent 6,506,559 and Massachusetts Institute of Technology and for laboratory and commercial research use only.
TrakMates is a registered trademark of Matrix Technologies Corporation.
© 2005 Sigma-Aldrich Co. Reproduction forbidden without permission.
Sigma-Aldrich brand products are sold exclusively through Sigma-Aldrich, Inc. Best viewed in IE6 or higher