|
Accuspin System Histopaque® 1077 |
 |
Hematology and Histology |
 |
 |
| |
|
Method: Density Gradient Centrifugation on Sodium DiatrizoatePolysucrose Gradients
Principle: Accuspin System Histopaque® 1077 employs centrifuge tubes specially designed with two chambers separated by a porous high-density polyethlyene barrier ("frit"). The lower chamber contains Histopaque® 1077 which allows the addition of anticoagulated whole blood without risk of mixing with the separation medium. On centrifugation, erythrocytes & granulocytes descend through the frit to pellet below the Histopaque® 1077. The erythrocytes aggregate, increasing their sedimentation rate, resulting in pelleting at the bottom of the Accuspin tube. Lymphocytes and other mononuclear cells, i.e., monocytes, remain above the frit at the plasma Histopaque® 1077 interface. |
|
Ordering Information
| Accuspin System-Histopaque-1077 | | |
| Product Description |
Product # |
Size/Quantity |
| Polypropylene radiation sterilized tube fitted with a high density polyethylene barrier and sterile filled with HISTOPAQUE®-1077. Packaged in a 12 mL centrifuge tube. Tube contains 3 ml HISTOPAQUE®-1077-1 | A 6929 | 40 x 3 mL |
| Polypropylene radiation sterilized tube fitted with a high density polyethylene barrier and sterile filled with HISTOPAQUE®-1077. Packaged in a 50 mL centrifuge tube. Tube contains 15 ml HISTOPAQUE®-1077-1 | A 7054 | 12 x 15 mL |
| Polypropylene radiation sterilized tube fitted with a high density polyethylene barrier and sterile filled with HISTOPAQUE®-1077. Packaged in a 50 mL centrifuge tube. Tube contains 15 ml HISTOPAQUE®-1077-1 | A 0561 | 100 x 15 mL |
| Accuspinä Tubes Nonsterile, 50 mL Capacity | A 1930 | 10 each |
| Accuspinä Tubes Sterile, 12 mL Capacity | A 1805 | 20 each |
| Accuspinä Tubes Sterile, 50 mL Capacity | A 2055 | 10 each |
| | | back to top | |