| BSC Certified Stains & Dyes |
back to top |
| Product Name | Product # |
| Acid Fuchsin calcium salt certified by the BSC for VanGieson connective tissue stain and Mallory aniline blue connective stain | A3908 |
| Alcian Blue 8GX certified | A3157 |
| Alizarin Red S certified by the BSC for (for gross staining of skeletons in small vertebrates and for differentiating bone from cartilage in mammalian embryos) | A5533 |
| Aniline Blue diammonium salt certified | 41,504-9 |
| Auramine O certified by the BSC for staining acid-fast bacteria in sputum or in paraffin sections of infected tissue. | A9655 |
| Azure A certified by the BSC for (for staining cell granules, microorganisms and other tissue. A constituent of Tetrachrome Stain (Macneal) for differentiating leucocytes.) | A2918 |
| Azure B certified by the BSC for differentiating cellular RNA and DNA in plant tissues. | A4043 |
| Basic Fuchsin certified | 85,734-3 |
| Basic Fuchsin certified by the BSC for staining bacterial flagella. | B0904 |
| Bismarck Brown B | 86,111-1 |
| Brilliant Cresyl Blue ALD certified by the BSC for staining the reticulum of immature red blood cells and platelets. | B5388 |
| Brilliant Green certified by the BSC for determining the presence of the colon organisms in drinking water and as a bacteriostatic agent. | B4014 |
| Carmine certified by the BSC for Orth's lithium carmine technique. | C6152 |
| Congo Red certified by the BSC for (for use with Mayer's hemalum as a cytoplasmic stain on tissues fixed in Bouin's fluid, and as a stain for amyloid in Highman's technique.) | C6277 |
| Cresyl Violet acetate certified by the BSC for the Powers and Clark method on formalin or Bouin's fixed spinal cord or brain. | C5042 |
| Crystal Violet certified by the BSC for the Gram stain; the Flemming triple stain with iodine; and Newton's crystal violet-iodine technique. | C0775 |
| Darrow Red certified | 21,188-5 |
| Eosin B Dye content min. 85 % | E8761 |
| Eosin Y disodium salt Dye content min. 80 % | E4382 |
| Erythrosin B sodium salt certified by the BSC for (for use with methylene blue as a plasma stain for nerve cells; for staining bacteria in soil; and as a phosphorescent triple probe for studying diffusion of membrane proteins.) | E8886 |
| Erythrosin extra bluish certified | 19,826-9 |
| Eosin alcohol soluble certified | 19,954-0 |
| Fast Green FCF certified by the BSC for use as a cytological counterstain and in Lillie's modification of the Masson's trichrome stain. | F7258 |
| Giemsa stain certified by the BSC for (for use in staining leukocytes in blood films and for staining malarial parasites in thin and thick films of blood.) | G5637 |
| Hematoxylin certified | H3136 |
| Indigo Carmine certified by the BSC for (for use with acid fuchsin as a stain for Negri bodies and with eosin Y as a counterstain to hematoxylins for staining vaginal smears.) | I2387 |
| Jenner's stain certified by the BSC for the Romanowsky-type stain for peripheral blood smears. | J1875 |
| Light Green SF Yellowish Dye content min. 65 % | L1886 |
| Malachite Green oxalate salt certified by the BSC for (for use as a counterstain on botanical material; as a spore stain for bacteria in the Schaeffer and Fulton method; and in Conklin's modification of the method.) | M9015 |
| Methyl Blue certified | M5528 |
| Methylene Blue certified by the BSC for histological and bacteriologic staining and as a constituent of Wright's blood stain. | M9140 |
| Methyl Orange certified | 23,410-9 |
| Methylene Violet (Bernthsen) certified | 21,631-3 |
| Methyl Green | M5015 |
| Methyl violet 2B certified | 19,809-9 |
| Neutral Red certified by the BSC for supravital staining of living blood cells. | N6634 |
| Nigrosin certified | 19,828-5 |
| Nile Blue A certified by the BSC for stain for fats and for the differentiation of melanins and lipofuscins. | N0766 |
| Oil Red O certified | 19,819-6 |
| Orange G certified by the BSC for Mallory's connective tissue stain and as a counterstain in histology and cytology. | O7252 |
| Orange II sodium salt certified by the BSC for (as a general tissue stain with eosin and azure C; with fast green FCF, safranin O and crystal violet in quadruple staining of tissues; as a stain for keratin in skin sections.) | O4505 |
| Orcein synthetic certified by the BSC for 8020 | O7505 |
| Pararosaniline hydrochloride Dye content min. 88 % | P1528 |
| Phloxine B certified by the BSC for Mallory's phloxine-methylene blue stain. | P4030 |
| Pyronin B certified 30 % (dye content) | 20,130-8 |
| Pyronin Y certified by the BSC for Kurnick's methyl green-pyronin stain on liver cells. | P6653 |
| Resazurin sodium salt certified | 19,930-3 |
| Rose Bengal sodium salt certified by the BSC for (as a fluorochrome for UV study of fats; for plate counts of soil fungi (bacteriostatic, but allows growth of fungi).) | R4507 |
| Safranin O certified by the BSC for (for use as a chromosome stain in the Flemming triple stain in combination with light green SF yellowish or fast green FCF; and in Gram stain.) | S8884 |
| Sudan III certified by the BSC for use as a fat stain. | S4136 |
| Sudan IV certified by the BSC for use as a fat stain. | S4261 |
| Sudan Black B certified | 19,966-4 |
| Thionin acetate Dye content min. 85 % | T7029 |
| Toluidine Blue O | T0394 |
| Wright stain certified by the BSC for staining blood and bone marrow films, and for staining malarial parasites in blood films. | W3000 |
|
Other Popular Stains & Dyes |
|
back to top |
| Product Name |
Grade |
Product # |
| 1-(2-Pyridylazo)-2-naphthol indicator grade |
Indicator |
101036 |
| 2-(5-Bromo-2-pyridylazo)-5-(diethylamino)phenol 97 % |
Technical |
180017 |
| 2,3,5-Triphenyltetrazolium chloride |
Technical |
T8877 |
| 2,6-Dichloroindophenol sodium salt hydrate |
Technical |
D1878 |
| 2',7'-Dichlorofluorescein ACS reagent |
ACS |
410217 |
| Chromotropic acid disodium salt dihydrate approx. 98 % |
Technical |
D5144 |
| 4-Amino-3-hydroxy-1-naphthalenesulfonic acid ACS reagent 98+ % ³98.0 % |
ACS |
398969 |
| Acid Fuchsin |
Technical |
F8129 |
| Acridine orange hydrochloride hydrate 99 % (HPLC) |
HPLC/TLC/
Electrophoresis |
318337 |
| Acridine orange solution 10 mg/ml in water |
Technical |
A8097 |
| Acridine orange hemi(zinc chloride) salt 90 % (dye content) |
Technical |
158550 |
| Adrenochrome |
Technical |
A5752 |
| Alcian Blue 8GX |
Technical |
236551 |
| Alcian Blue 8GX powder |
Technical |
A5268 |
| Alcian Blue, pyridine variant |
Technical |
335630 |
| Alcian Blue-tetrakis(methylpyridinium) chloride |
Technical |
A4045 |
| Alizarin |
Technical |
122777 |
| Alizarin yellow R |
Technical |
195073 |
| Allura Red AC |
Technical |
458848 |
| Aurintricarboxylic acid ammonium salt ACS reagent |
ACS |
A36883 |
| Amaranth |
Technical |
A1016 |
| Anthrone |
Technical |
A1631 |
| Anthrone ACS reagent 97 % |
ACS |
319899 |
| Arsenazo III |
Technical |
A92775 |
| Astra Blue |
Technical |
363405 |
| Auramine O 80 % (dye content) |
Technical |
856533 |
| Azobenzene 99 % |
Technical |
424633 |
| Azure B |
Technical |
227935 |
| Azure II A mixture of Azure B and Methylene Blue in equal amounts. |
Technical |
861065 |
| Basic Fuchsin |
Technical |
215597 |
| Benzophenone imine 97 % |
Technical |
293733 |
| Biotin-4-Fluorescein |
Technical |
B9431 |
| Bisbenzimide H 33258 min. 98 % (TLC) |
Technical |
B2883 |
| Bismarck Brown Y (G) |
Technical |
B2759 |
| Brilliant Black BN |
Technical |
211842 |
| Ponceau BS |
Technical |
B6008 |
| Brilliant Blue G Dye content approx. 60 % by elemental analysis |
Technical |
B1131 |
| Brilliant Blue G min. 90 % (TLC) |
HPLC/TLC/
Electrophoresis |
B5133 |
| Brilliant Blue G solution Concentrate |
Technical |
B8522 |
| Brilliant Blue G Dye content approx. 90 % by elemental analysis |
Technical |
B0770 |
| Brilliant Blue R Dye content approx. 50 % |
Technical |
B0149 |
| Brilliant Blue R Dye content approx. 90 % |
Technical |
B7920 |
| Brilliant Blue R Concentrate |
Technical |
B8647 |
| Brilliant Blue R Staining Solution Ethanol based |
Technical |
B6529 |
| PhastGel Blue R |
Technical |
B4921 |
| Brilliant Cresyl Blue ALD |
Technical |
860867 |
| Brilliant Green |
Technical |
B6756 |
| Bromocresol Green Sultone Form |
Technical |
B0381 |
| Bromocresol Green sodium salt crystalline |
Technical |
B1256 |
| Bromocresol Green sodium salt ACS reagent |
ACS |
114367 |
| Bromocresol Purple |
Technical |
B5880 |
| Bromobimane min. 97 % |
Technical |
B4380 |
| Bromocresol Green sodium salt solution 0.04 wt. % in water |
Technical |
318701 |
| Bromocresol Green Sultone Form ACS reagent |
ACS |
114359 |
| Bromocresol Purple indicator grade |
Indicator |
114375 |
| Bromocresol Purple solution 0.04 wt. % in water |
Technical |
318736 |
| Bromocresol Purple sodium salt indicator grade |
Indicator |
860891 |
| Bromophenol Blue solution 0.04 wt. % in water |
Technical |
318744 |
| Bromphenol Blue sodium salt ACS reagent |
ACS |
114405 |
| Bromophenol Red sodium salt |
Technical |
B7010 |
| Bromosulfalein |
Technical |
S0252 |
| Bromphenol Blue sodium salt |
Technical |
B6131 |
| Bromphenol Blue sodium salt for molecular biology, for electrophoresis |
HPLC/TLC/
Electrophoresis |
B8026 |
| Bromphenol Blue sodium salt for electrophoresis |
HPLC/TLC/
Electrophoresis |
B5525 |
| Bromophenol Blue |
Technical |
B0126 |
| Bromothymol blue sodium salt |
Technical |
B8630 |
| Bromothymol blue sodium salt ACS reagent |
ACS |
114421 |
| Calcein |
Technical |
C0875 |
| Calcein AM solution min. 4 mM in DMSO min. 90 % (TLC) liquid |
Technical |
C1359 |
| Calcon® |
Technical |
H1878 |
| Calmagite indicator grade |
Indicator |
C204 |
| Canada balsam |
Technical |
C1795 |
| Carbazole approx. 95 % |
Technical |
C5132 |
| Carbol Fuchsin |
Technical |
C4165 |
| Carbostyril 124 99 % |
Technical |
363308 |
| Cardiogreen |
Technical |
I2633 |
| Carmine powder |
Technical |
C1022 |
| Carminic acid |
Technical |
229253 |
| Celestine blue |
Technical |
206342 |
| Chicago Sky Blue 6B powder |
Technical |
C8679 |
| Chlorophenol Red indicator grade |
Indicator |
199524 |
| Chlorophenol Red sodium salt |
Technical |
235482 |
| Chlorophyll a from Anacystis nidulans algae |
Technical |
C6144 |
| Chlorophyll b from spinach |
Technical |
C5878 |
| Chromeazurol B |
Technical |
M1782 |
| Chromeazurol S |
Technical |
199532 |
| Chromotrope 2R suitable for modified Gomori Trichrome stain |
Technical |
C3143 |
| Chromotrope 2R |
Technical |
202088 |
| Chromotropic acid disodium salt dihydrate ACS reagent |
ACS |
213276 |
| Chrysin |
Technical |
C80105 |
| Cibacron Blue 3G-A |
Technical |
C9534 |
| Congo Red |
Technical |
C6767 |
| Coumarin 6 98 % |
Technical |
442631 |
| Coumarin 314 |
Technical |
392995 |
| Coumarin 343 |
Technical |
393029 |
| Practice of Thin Layer Chromatography, 3rd ed. |
Technical |
P5691 |
| m-Cresol Purple sodium salt |
Technical |
211761 |
| Cresol red indicator grade |
Indicator |
114472 |
| Cresol Red sodium salt indicator grade |
Indicator |
114480 |
| Cresyl Violet acetate |
Technical |
C1791 |
| Crocein Scarlet 7B |
Technical |
C8822 |
| Crystal Violet Dye content approx. 90 % |
Technical |
C3886 |
| Crystal Violet ACS reagent ³90.0 % as anhydrous |
ACS |
C6158 |
| Curcumin from Curcuma longa (Turmeric) 65-70 % (typically) powder |
Technical |
C1386 |
| Phloxine B crystalline |
Technical |
P2759 |
| Demecolcine min. 98 % (TLC) |
Technical |
D7385 |
| Diamidino yellow dihydrochloride |
Technical |
D0281 |
| o-Dianisidine dihydrochloride tech. |
Technical |
191248 |
| o-Dianisidine dihydrochloride crystalline |
Technical |
D3252 |
| o-Dianisidine reagent |
Technical |
D2679 |
| Dihydroethidium |
Technical |
D7008 |
| Dimidium bromide 95 % |
Technical |
234281 |
| Diphenylamine |
Technical |
D2385 |
| Dithizone |
Technical |
D5130 |
| Dithizone ³85.0 % ACS reagent |
ACS |
194832 |
| Direct Blue 71 |
Technical |
212407 |
| Direct Red 80 |
Technical |
365548 |
| Dithizone |
Technical |
D5130 |
| Dithizone ³85.0 % ACS reagent |
ACS |
194832 |
| Eosin Y Dye content approx. 99 % |
Technical |
E4009 |
| Eosin Y disodium salt Dye content approx. 85 % |
Technical |
E6003 |
| Eosin Y solution 5 wt. % in water |
Technical |
318906 |
| Eriochrome Black T ACS reagent, indicator grade |
ACS |
858390 |
| Eriochrome Cyanine R practical grade |
Technical |
E2502 |
| Erythrosin B 95+ % |
Technical |
200964 |
| Ethidium bromide approx. 95 % (HPLC) |
Technical |
E8751 |
| Ethidium homodimer solution |
Technical |
E1903 |
| Ethyl Orange sodium salt indicator grade |
Indicator |
199567 |
| Evans Blue |
Technical |
E2129 |
| Fast Black K Salt hemi(zinc chloride) salt practical grade |
Technical |
F7253 |
| Fast Blue BB Salt hemi(zinc chloride) salt |
Technical |
F3378 |
| Fast Blue B Salt |
Technical |
340774 |
| Fast Corinth V zinc chloride double salt |
Technical |
227366 |
| Fast Garnet GBC sulfate salt |
Technical |
F8761 |
| Fast Green FCF Dye content approx. 90 % |
Technical |
F7252 |
| Fast Red Violet LB Salt |
Technical |
F3381 |
| Fast Violet B Salt crystalline |
Technical |
F1631 |
| Ferroin solution 0.1 wt. % in water |
Technical |
318922 |
| Fluo 3-AM approx. 70 % |
Technical |
F6142 |
| Fluorescamine 98 % |
Technical |
201650 |
| Fluorescein sodium salt |
Technical |
F6377 |
| Fluoresceinamine isomer I |
Technical |
201626 |
| Fluorescein diacetate |
Technical |
F7378 |
| Fluorescein 5(6)-isothiocyanate approx. 90 % (HPLC) |
Technical |
F3651 |
| Fluorescein isothiocyanate isomer I suitable for protein labeling min. 90 % (HPLC) powder |
Technical |
F7250 |
| Fluorescein isothiocyanate isomer I approx. 98 % (HPLC) |
Technical |
F4274 |
| Fluorescein isothiocyanate isomer I 90 % |
Technical |
F2502 |
| Fluorescent Brightener 28 |
Technical |
F3543 |
| Fura 2-AM powder |
Technical |
F0888 |
| Fura PE 3-AM approx. 90 % |
Technical |
F0918 |
| Fusidic acid |
Technical |
F0756 |
| Gentian Violet USP |
USP |
G2039 |
| Hematoxylin |
Technical |
H9627 |
| Hydroxynaphthol Blue disodium salt ACS reagent |
ACS |
219916 |
| Indigo Carmine |
Technical |
I8130 |
| Iodonitrotetrazolium chloride |
Technical |
I8377 |
| Janus Green B |
Technical |
J1000 |
| Jenner's stain suitable for blood stain |
Technical |
J1625 |
| Keratin azure |
Technical |
K8500 |
| Leishman's stain |
Technical |
L6254 |
| Light Green SF Yellowish crystalline |
Technical |
L5382 |
| Lucifer Yellow CH dilithium salt |
Technical |
L0259 |
| Lucifer Yellow CH dipotassium salt 90 % (dye content) |
Technical |
328197 |
| Lumichrome |
Technical |
103217 |
| Malachite Green oxalate salt |
Technical |
M6880 |
| Malachite Green Carbinol base |
Technical |
229105 |
| Melanin from Sepia officinalis |
Technical |
M2649 |
| Metanil Yellow |
Technical |
202029 |
| Methyl 3-amino-5,6-dichloro-2-pyrazinecarboxylate 97 % |
Technical |
277878 |
| Methyl Blue |
Technical |
M6900 |
| Methyl Green approx. 85 % |
Technical |
M8884 |
| Methyl Orange ACS reagent |
ACS |
114510 |
| Methyl Purple in solution in water |
Technical |
277339 |
| Methyl Red |
Technical |
M7267 |
| Methyl Red hydrochloride ACS reagent |
ACS |
861235 |
| Methyl Red sodium salt |
Technical |
M8256 |
| Methyl Red crystals ACS reagent |
ACS |
250198 |
| Methyl Red sodium salt ACS reagent |
ACS |
114502 |
| Methylene Blue |
Technical |
M44907 |
| Methylene Blue solution |
Technical |
1808 |
| Methylene Blue |
Technical |
MB1 |
| Methylene Blue |
Cell/Molecular |
M4159 |
| Methylene Violet 3RAX |
Technical |
307505 |
| Methylthymol Blue sodium salt |
Technical |
B42004 |
| MTT Formazan powder |
Technical |
M2003 |
| Murexide ACS reagent |
ACS |
222461 |
| Naphthol Blue Black |
Technical |
195243 |
| Naphthol Green B |
Technical |
N7257 |
| Naphthol Yellow S |
Technical |
199621 |
| a-Naphtholphthalein practical grade |
Technical |
N8257 |
| N,N-Diethyl-p-phenylenediamine hemioxalate salt |
Technical |
D5143 |
| Neutral Red powder cell culture tested |
Cell/Molecular |
N4638 |
| Neutral Red practical grade |
Technical |
N2880 |
| Neutral Red |
Technical |
N7005 |
| New Fuchsin |
Technical |
N0638 |
| Nigrosin water soluble |
Technical |
N4754 |
| Nile Red |
Technical |
N3013 |
| Ninhydrin 97 % |
Technical |
454044 |
| Nitro Blue Tetrazolium tablet |
Cell/Molecular |
N5514 |
| Nitrotetrazolium blue chloride approx. 98 % (TLC) |
HPLC/TLC/
Electrophoresis |
N6876 |
| Nitrotetrazolium blue chloride powder |
Cell/Molecular |
N6639 |
| Nitrotetrazolium Blue chloride monohydrate |
Technical |
N15405 |
| Nuclear Fast Red suitable for determination of calcium |
Technical |
N8002 |
| Oil Red O powder |
Technical |
O0625 |
| Orange G |
Technical |
O1625 |
| Orange G |
Cell/Molecular |
O3756 |
| Orcein synthetic |
Technical |
O7380 |
| Orcinol monohydrate from lichens |
Technical |
O1875 |
| Osmium tetroxide |
Technical |
O5500 |
| Oxacillin sodium salt hydrate |
Technical |
O1002 |
| Pararosaniline Base crystalline |
Technical |
P7632 |
| Pararosaniline hydrochloride powder |
Technical |
P3750 |
| Perylene Purified by sublimation 99.5+ % |
Technical |
394475 |
| Perylene 99+ % |
Technical |
P11204 |
| PhastGel Blue R |
Technical |
B4921 |
| Phenazine ethosulfate approx. 95 % |
Technical |
P4544 |
| Phenol red solution liquid 0.5% cell culture tested |
Cell/Molecular |
P0290 |
| Phenol red solution for electrophoresis |
HPLC/TLC/
Electrophoresis |
P8421 |
| Phenol Red |
Technical |
P4633 |
| Phenol Red powder cell culture tested |
Cell/Molecular | |