|
2-Propanol Physical Properties |
 |
2-Propanol |
 |
 |
| |
|
| Sigma-Aldrich 2-Propanol |
| Synonym |
Isopropanol |
Beilstein Registry |
635639 |
| Molecular Formula |
(CH3)2CHOH |
EG/EC Number |
2006617 |
| Molecular Weight |
60.10 |
MDL Number |
MFCD00011674 |
| CAS Number |
67-63-0 |
Vapor Pressure |
33 mm Hg (20°C) |
| Density |
0.785 g/mL at 25°C |
Dielectric Constant |
20.18 at 20°C |
| Boiling Point |
82°C |
Dipole Moment |
1.560 |
| Melting Point |
-89.5°C |
UV cutoff |
210 nm |
| Flash Point |
11.7°C |
Refractive Index |
1.377 at 20°C |
| Viscosity |
2.040 at 25°C |
Vapor Density |
2.1 (vs. air) |
| RIDADR |
UN 1219 3/PG 2 |
Risk Statements |
11-36-67 |
| Hazard Codes |
F,Xi |
Safety Statements |
7-16-24/25-26 |
 |
2-Propanol Specification Comparison Table