|
Acetone Physical Properties |
 |
Acetone |
 |
 |
| |
|
| Sigma-Aldrich Acetone |
| Synonym |
2-Propanone |
Beilstein Registry |
635680 |
| Molecular Formula |
(CH3)COCH3 |
EG/EC Number |
2006622 |
| Molecular Weight |
58.08 |
MDL Number |
MFCD00008765 |
| CAS Number |
67-64-1 |
Vapor Pressure |
184
mm Hg (20°C) |
| Density |
0.791
g/mL at 25ēC |
Dielectric Constant |
21.01
at 20ēC |
| Boiling Point |
56ēC |
Dipole Moment |
2.880 |
| Melting Point |
-
94ēC |
UV cutoff |
330 nm |
| Flash Point |
-17.2ēC |
Refractive Index |
1.359
at 20ēC |
| Viscosity |
0.306
at 25ēC |
Vapor Density |
2 (vs. air) |
| RIDADR |
UN 1090 3/PG 2 |
Risk Statements |
11-36-66-67 |
| Hazard Codes |
F,Xi |
Safety Statements |
9-16-26 |
 |
Acetone Specification Comparison Table