|
Acetonitrile Physical Properties |
 |
Acetonitrile |
 |
 |
| |
|
| Sigma-Aldrich Acetonitrile |
| Synonym |
Methyl cyanide |
Beilstein Registry |
741857 |
| Molecular Formula |
CH3CN |
EG/EC Number |
2008352 |
| Molecular Weight |
41.05 |
MDL Number |
MFCD00001878 |
| CAS Number |
75-05-8 |
Vapor Pressure |
72.8
mm Hg (20°C) |
| Density |
0.786
g/mL at 25ēC |
Dielectric Constant |
36.64
at 20ēC |
| Boiling Point |
81-82ēC |
Dipole Moment |
3.924 |
| Melting Point |
-48ēC |
UV cutoff |
190
nm |
| Flash Point |
5.5ēC |
Refractive Index |
1.344
at 20ēC |
| Viscosity |
0.369
at 25ēC |
Vapor Density |
1.41 (vs. air) |
| RIDADR |
UN 1648 3/PG 2 |
Risk Statements |
11-20/21/22-36 |
| Hazard Codes |
F,Xn |
Safety Statements |
16-36/37 |
 |
Acetonitrile Specification Comparison Table