| Sigma-Aldrich Acetonitrile Specifications Comparison |
| |
CHROMASOLV
LC-MS |
CHROMASOLV
Plus |
CHROMASOLV
Gradient |
Biotech |
Anhydrous |
Spectro-
photometric |
ACS
|
ReagentPlus |
| Purity |
≥99.9% |
≥99.9% |
≥99.9% |
99.93% |
99.8% |
≥99.5% |
≥99.5% |
99% |
| UV Absorption |
| λ: 190 nm |
|
|
|
≤1.00 |
|
| λ: 193 nm |
|
≤1.00 |
|
|
| λ: 195 nm |
≤0.097 |
≤0.10 |
≤0.13 |
|
| λ: 200 nm |
≤0.022 |
≤0.02 |
≤0.03 |
|
| λ: 210 nm |
|
|
|
≤0.03 |
|
≤0.30 |
|
|
| λ: 220 nm |
|
|
|
≤0.01
|
|
≤0.10 |
|
|
| λ: 228 nm |
|
≤0.005 |
≤0.004 |
|
| λ: 230 nm |
≤0.0044 |
|
| λ: 235 nm |
|
|
|
≤0.005 |
|
≤0.02 |
|
|
| λ: 250 nm |
|
≤0.01 |
|
|
| λ: 400 nm |
|
|
|
≤0.005 |
|
≤0.01 |
|
|
| Gradient analysis |
| 210 nm |
≤1 mAU |
≤1 mAU |
≤3 mAU |
|
| 254 nm |
≤0.2 mAU |
≤0.2 mAU |
≤0.5 mAU |
|
| Total Impurities |
| Water |
≤0.01% |
≤0.01% |
≤0.02% |
≤0.001% |
≤0.001% |
≤0.03% |
≤0.3% |
|
| |
|
|
|
≤0.005% (100mL) |
|
|
|
| Evaporation Residue |
≤0.0002% |
≤0.0002% |
≤0.0005% |
≤0.0002% |
≤0.0005% |
≤0.0003% |
≤0.005% |
|
| Halogenated Residue |
|
10 ng/L (as Heptachlor epoxide) |
|
| Fluorescence 254 nm |
≤0.5 ppb |
≤0.5 ppb |
≤1.0ppb |
|
| Fluorescence 365 nm |
≤0.5 ppb |
≤0.5 ppb |
≤0.5 ppb |
|
| LC-MS suitability |
passes test |
|
| Alkalinity |
≤0.0002% (NH3) |
≤0.0001% (NH3) |
≤0.0002% (NH3) |
|
| Titratable acid |
≤0.001% free acid (as CH3COOH) |
≤0.001% free acid (as CH3COOH) |
≤0.001% free acid (as CH3COOH) |
<0.008 meq/g |
|
|
<0.008 meq/g |
|
| Titratable base |
|
|
|
<0.0006 meq/g |
|
|
<0.0006 meq/g |
|
| Silver |
0.1 ppm |
|
| Aluminum |
0.5 ppm |
|
| Barium |
0.1 ppm |
|
| Calcium |
0.1 ppm |
|
| Cadmium |
0.05 ppm |
|
| Cobalt |
0.02 ppm |
|
| Chromium |
0.02 ppm |
|
| Copper |
0.01 ppm |
|
| Iron |
0.1 ppm |
|
| Potassium |
0.1 ppm |
|
| Magnesium |
0.1 ppm |
|
| Manganese |
0.01 ppm |
|
| Sodium |
0.1 ppm |
|
| Nickel |
0.02 ppm |
|
| Lead |
0.1 ppm |
|
| Tin |
0.1 ppm |
|
| Zinc |
0.1 ppm |
|
| Packaging Options |
| |
CHROMASOLV
LC-MS |
CHROMASOLV
Plus |
CHROMASOLV
Gradient |
Biotech |
Anhydrous |
Spectro-
photometric |
ACS
|
ReagentPlus |
| Glass Bottle |
34967 |
34998 |
34851 |
|
|
154601 |
360457 |
110086 |
| PVC Coated Bottle |
|
675415 |
439134 |
|
|
|
437557 |
|
| Sure/Seal |
|
|
|
494445 |
271004 |
|
|
|
| VerSA-Flow |
|
|
439134 |
|
| NOWPAK |
|
34998 |
439134 |
|
| Pure-Pac |
|
34998 |
34851 |
494445 |
271004 |
|
|
|
| Carbon Steel |
|
360457 |
|
| Drum |
|
|
439134 |
|
|
|
360457 |
|
For a complete list of all available unit sizes and product specifications,
simply click on the product numbers above!
CHROMASOLV® Mobile Phase Blends for HPLC & LC-MS Applications