Sigma-Aldrich's offers a full range of high-purity solvents with extremely low water levels specifically manufactured for moisture sensitive Organic and BioTech applications.
The BioTech line of solvents is characterized by not only low water content, but also minimal residue and clean UV spectra.
| Anhydrous |
Biotech |
| Acetonitrile |
Purity Water 100 mL Residue |
27,100-4 99.8% < 10 ppm < 50 ppm < 0.0005% |
|
49,444-5 ≥99.93% < 10 ppm <0.0002% |
UV nm 190 210 220 235-400 <0.008 meq/g <0.0006 meq/g |
Max Abs. 1.000 0.030 0.010 0.005 Titr. Acid Titr. Base |
| Buytl acetate |
Purity Water Residue |
28,772-5 ≥99% < 50 ppm < 0.0005% |
|
49,616-2 99.7% < 10 ppm <0.0005% |
UV nm
<0.0016 meq/g |
Max Abs.
Titr. Acid |
| Carbon tetrachloride |
Purity Water 100 mL Residue |
28,911-6 ≥99.5% < 20 ppm <50 ppm < 0.0003% |
|
| 1-Chlorobutane |
Purity Water 100 mL Residue |
41,425-5 99.5% < 30 ppm < 50 ppm < 0.0005% |
|
49,617-0 ≥99.5% < 10 ppm
<0.0003% |
UV nm
<0.0016 meq/g |
Max Abs.
Titr. Acid |
Chloroform 0.5-1.0% ethanol as stabilizer |
Purity Water 100 mL Residue |
40,176-5 99.7% < 10 ppm < 50 ppm < 0.0005% |
|
49,618-9 ≥99.9% < 10 ppm
<0.0003% |
|
|
Dichloromethane 50-150 ppm hydrocarbon as stabilizer |
Purity Water 100 mL Residue |
27,099-7 99.0% < 10 ppm < 50 ppm < 0.0005% |
|
49,445-3 99.9% < 20 ppm
<0.0003% |
UV nm 235 240 250 260 340-400 |
Max Abs. 1.000 0.200 0.050 0.020 0.010 |
| N,N-Diisopropylethylamine |
Purity Water Residue |
27,101-2 99.8% < 50 ppm < 0.0001% |
|
49,621-9 99.5% < 20 ppm |
|
|
| N,N-Dimethylformamide |
Purity Water Residue |
22,705-6 99.8% < 50 ppm < 0.0005% |
|
49,448-8 ≥99.9% < 50 ppm <0.0005% |
UV nm 275 295 310 340-400 <0.0005 meq/g <0.0003 meq/g |
Max Abs. 0.30 0.10 0.05 0.07 Titr. Acid Titr. Base |
| Dimethyl sulfoxide |
Purity Water Residue |
27,685-5 ≥99.9% < 50 ppm < 0.0005% |
|
49,442-9 99.8% < 50 ppm <0.0005% |
UV nm 270 280 295 350-400 |
Max Abs. 1.00 0.30 0.10 0.01 |
| 1,4-Dioxane |
Purity Water 100 mL Residue |
29,630-9 99.8% < 30 ppm < 50 ppm < 0.0003% |
|
Ether 1 ppm BHT as stabilzer |
Purity Water 100 mL Residue |
29,608-2 ≥ 99.7% < 30 ppm < 50 ppm < 0.0003% |
|
| Ethyl actate |
Purity Water Residue |
27,098-9 99.8% < 50 ppm < 0.0005% |
|
49,451-8 ≥99.8% < 10 ppm <0.0003% |
UV nm 254 263 275-400 <0.0009 meq/g |
Max Abs. 1.00 0.05 0.01 Titr. Acid |
| Heptane |
Purity Water 100 mL Residue |
24,665-4 99% < 10 ppm < 50 ppm < 0.0005% |
|
49,452-6 ≥99% < 50 ppm
<0.0002% |
UV nm 197 210 220 230 260-400 |
Max Abs. 1.00 0.40 0.15 0.05 0.01 |
| Methanol |
Purity Water 100 mL Residue |
32,241-5 99.8% < 20 ppm < 50 ppm < 0.0003% |
|
49,443-7 99.93% < 20 ppm
<0.0003% |
UV nm 203 220 230 240 260-400 <0.0003 meq/g <0.0002 meq/g |
Max Abs. 1.00 0.60 0.15 0.05 0.01 Titr. Acid Titr. Base |
| 3-Methyl-1-butanol |
Purity Water 100 mL Residue |
30,943-5 ≥99% < 30 ppm < 50 ppm < 0.0005% |
|
49,620-0 ≥99% < 50 ppm
<0.0005% |
UV nm
<0.002 meq/g <0.1% |
Max Abs.
Titr. Acid Carbonyl |
| 1-Methyl-2-pyrrolidinone |
Purity Water |
32,863-4 99.5% < 50 ppm |
|
49,449-6 ≥99.5% < 50 ppm |
UV nm 285 300 325 350-400 <0.01% meq/g |
Max Abs. 1.00 0.50 0.10 0.01 Free amines |
| 1-Propanol |
Purity Water Residue |
27,954-4 99.7% < 50 ppm < 0.0003% |
|
49,619-7 99.7% <0.0003% |
UV nm
<0.0004 meq/g |
Max Abs.
Titr. Acid |
| Pyridine |
Purity Water 100 mL Residue |
27,097-0 99.8% < 30 ppm < 50 ppm < 0.0005% |
|
49,441-0 ≥99.9% < 30 ppm
<0.0003% |
UV nm <0.001% <0.0005% <0.002% <0.0001% |
Max Abs. Cl Cu NH3 SO4 |
Tetrahydrofuran Inhibitor-free |
Purity Water 100 mL Residue |
401757 99.9% < 20 ppm < 50 ppm < 0.0005% |
|
49,446-1 ≥99.9% < 50 ppm
<0.0005% |
UV nm
<0.005% |
Max Abs.
Peroxides |
Tetrahydrofuran 250 ppm BHT as stabilizer |
Purity Water 100 mL |
186562 99.9% < 20 ppm < 50 ppm |
|
| Toluene |
Purity Water 100 mL Residue |
24,451-1 99.8% < 10 ppm < 50 ppm <0.0005% |