Order Center
Research Essentials
Biological Buffers
Hematology and Histology
Solvents
2-Propanol Center
Acetone Center
Acetonitrile Center
Anhydrous BioTech
Bottles and Cans
Aluminum Bottles
Amber Glass Bottles
PVC Coated Bottles
PVC SureSeal
Sure Seal
CHROMASOLV
Chloroform Center
Dichloromethane Center
Dimethyl Sulfoxide Center
Ethyl Acetate Center
General Reagents
Hexane Center
High-Purity
Methanol Center
Semi Bulk
Tetrahydrofuran Center
Toluene Center
Water Center
Key Resources
Virtual Lab
Biochemicals
Chemicals
Key Resources
 Solvents
Aluminum Bottles
These one-piece seamless aluminum bottles are corrosion resistant and U.N. Approved.
  • Pure 1050A aluminum (min. 99.5%)
  • Polypropylene closure with PTFE liner
  • Anti-back off, tamper evident locking closure
  • Easily recyclable
Products available in Aluminum Bottles
Capacity Bottle Dimension (H x D) cm Closure Case Pack Case Pack Dimension (L x W x H) cm
1 L 24.3 x 8.8 32 mm 6 x 1 L 39.37 x 26.83 x 27.94
4 L 23.9 x 17.5 45 mm 4 x 4 L 36.20 x 36.20 x 39.37