Order Center
Research Essentials
Biological Buffers
Hematology and Histology
Solvents
2-Propanol Center
Acetone Center
Acetonitrile Center
Anhydrous BioTech
Bottles and Cans
CHROMASOLV
Chloroform Center
Amylene Specification
Ethanol Specification
Miscibility / Immiscibility
Physical Properties
With UV Specification
Without UV Specification
Dichloromethane Center
Dimethyl Sulfoxide Center
Ethyl Acetate Center
General Reagents
Hexane Center
High-Purity
Methanol Center
Semi Bulk
Tetrahydrofuran Center
Toluene Center
Water Center
Key Resources
Virtual Lab
Biochemicals
Chemicals
Key Resources
 Chloroform with UV Specification Comparison
Chloroform with UV Specifications
Chloroform With UV Specifications Specification Comparison Printer friendly PDF version (123 kb)
Sigma-Aldrich Chloroform Specifications Comparison
With UV Specifications
  CHROMASOLV
Plus
CHROMASOLV
Plus
CHROMASOLV
for HPLC
CHROMASOLV
for HPLC
ACS
Spectro-
photometric
ACS
Spectro-
photometric
Purity ≥99.9% ≥99.9% ≥99.8% ≥99.8% ≥99.8% ≥99.8%
UV Absorption
λ: 245 nm ≤1.00 ≤1.00 ≤1.00   ≤1.00 ≤1.00
λ: 250 nm ≤0.30 ≤0.30   ≤1.00
λ: 255 nm     ≤0.15   ≤0.25 ≤0.25
λ: 260 nm     ≤0.15 ≤0.30 ≤0.15 ≤0.15
λ: 270 nm     ≤0.02   ≤0.05 ≤0.05
λ: 275 nm ≤0.005 ≤0.005        
λ: 280 nm       ≤0.005    
λ: 290 nm     ≤0.01   ≤0.01 ≤0.01
λ: 400 nm ≤0.005 ≤0.005 ≤0.01   ≤0.01 ≤0.01
Total Impurities
Stabilizer 0.5-1.0% Ethanol Amylenes 0.5-1.0% Ethanol Amylenes 0.5-1.0% Ethanol Amylenes
Water ≤0.02% ≤0.02% ≤0.01% ≤0.01% ≤0.03% ≤0.03%
Evaporation Residue ≤0.0001% ≤0.0001% ≤0.0003% ≤0.0005% ≤0.001% ≤0.001%
Halogenated Residue ≤10 ng/L (as Heptachlor epoxide) ≤10 ng/L (as Heptachlor epoxide)    
Fluorescence 365 nm ≤0.5 ppb ≤0.5 ppb        
Acetaldehyde     Passes Test   Passes Test Passes Test
Acetone     Passes Test   Passes Test Passes Test
Titratable acid       ≤0.001% free acid (as HCL) Passes Test  
Free Chlorine     Passes Test   Passes Test Passes Test
Formaldehyde   Passes Test     Passes Test Passes Test
Lead     ≤0.05 ppm   ≤0.05 ppm ≤0.05 ppm
Packaging Options
  CHROMASOLV
Plus
CHROMASOLV
Plus
CHROMASOLV
for HPLC
CHROMASOLV
for HPLC
ACS
Spectro-
photometric
ACS
Spectro-
photometric
Glass Bottle 650471 650498 366927 34854 366919 154733
PVC Coated Bottle     439142      
NOWPAK     34854    
For a complete list of all available unit sizes and product specifications,
simply click on the product numbers above!