Order Center
Research Essentials
Biological Buffers
Hematology and Histology
Solvents
2-Propanol Center
Acetone Center
Acetonitrile Center
Anhydrous BioTech
Bottles and Cans
CHROMASOLV
Chloroform Center
Amylene Specification
Ethanol Specification
Miscibility / Immiscibility
Physical Properties
With UV Specification
Without UV Specification
Dichloromethane Center
Dimethyl Sulfoxide Center
Ethyl Acetate Center
General Reagents
Hexane Center
High-Purity
Methanol Center
Semi Bulk
Tetrahydrofuran Center
Toluene Center
Water Center
Key Resources
Virtual Lab
Biochemicals
Chemicals
Key Resources
 Chloroform without UV Specification Comparison
Chloroform Without UV Specifications
Chloroform Without UV Specifications Specification Comparison Printer friendly PDF version (74 kb)
Sigma-Aldrich Chloroform Specifications Comparison
Without UV Specifications
  Biotech Anhydrous Anhydrous ACS ACS ReagentPlus
Purity ≥99.8% ≥99% ≥99% ≥99.8% ≥99.8% ≥99.8%
Total Impurities
Stabilizer 0.5-1.0% Ethanol 0.5-1.0% Ethanol Amylenes 0.5-1.0% Ethanol Amylenes 0.5-1.0% Ethanol
Water ≤0.01% ≤0.001% ≤0.001%   ≤0.03% ≤0.03%
  ≤0.005% (100mL) ≤0.005% (100mL)      
Evaporation Residue ≤0.0003% ≤0.0003% ≤0.0003% ≤0.001% ≤0.001%  
Acetaldehyde       Passes Test Passes Test  
Acetone       Passes Test Passes Test  
Titratable acid       Passes Test    
Free Chlorine Passes Test     Passes Test Passes Test  
Formaldehyde       Passes Test Passes Test  
Lead ≤0.05 ppm     ≤0.05 ppm ≤0.05 ppm  
Packaging Options
  Biotech Anhydrous Anhydrous ACS ACS ReagentPlus
Glass Bottle   288306   319988 472476 132950
PVC Coated Bottle       437581 480150 320242
Sure/Seal 496189 372978      
Aluminum Bottle     676888 676896  
Pure-Pac 496189 372978      
Carbon Steel     319988   132950
For a complete list of all available unit sizes and product specifications,
simply click on the product numbers above!