|
Dichloromethane Physical Properties |
 |
Dichloromethane |
 |
 |
| |
|
| Sigma-Aldrich
Dichloromethane
|
| Synonym |
Methylene chloride |
Beilstein Registry |
1730800 |
| Molecular Formula |
CH2Cl2
|
EG/EC Number |
2008389 |
| Molecular Weight |
84.93 |
MDL Number |
MFCD00000881 |
| CAS Number |
75-09-2 |
Vapor Pressure |
353
mm Hg (20°C) |
| Density |
1.325
g/mL at 25ēC |
Dielectric Constant |
8.93
at 25ēC |
| Boiling Point |
39.8-40.0ēC |
Dipole Moment |
1.600 |
| Melting Point |
-97ēC |
UV cutoff |
235
nm |
| Flash Point |
- |
Refractive Index |
1.424
at 20ēC |
| Viscosity |
0.413
at 25ēC |
Vapor Density |
2.9 (vs. air) |
| RIDADR |
UN 1593 6.1/PG 3 |
Risk Statements |
40 |
| Hazard Codes |
Xn |
Safety Statements |
23-24/25-36/37 |
 |
Dichloromethane Specification Comparison Table