Sigma-Aldrich Dichloromethane Specifications Comparison
|
| |
CHROMASOLV
Plus |
CHROMASOLV
for HPLC |
Biotech |
Anhydrous |
ACS
Spectro-
photometric |
ACS |
| Purity |
≥99.9% |
≥99.8% |
99.9% |
≥99.8% |
≥99.5% |
≥99.5% |
| UV Absorption |
| λ: 235 nm |
≤1.00 |
≤0.70 |
≤1.00 |
|
≤1.00 |
|
| λ: 240 nm |
≤0.10 |
≤0.30 |
≤0.20 |
|
≤0.35 |
|
| λ: 245 nm |
|
≤0.10 |
|
| λ: 250 nm |
≤0.02 |
|
≤0.05 |
|
≤0.10 |
|
| λ: 260 nm |
|
≤0.01 |
≤0.02 |
|
≤0.04 |
|
| λ: 300 nm |
≤0.005 |
|
| λ: 340 nm |
|
|
≤0.01 |
|
≤0.01 |
|
| λ: 400 nm |
≤0.005 |
|
≤0.01 |
|
≤0.01 |
|
| Total Impurities |
| Stabilizer |
50-150 ppm Amylene |
50-150 ppm Amylene |
50-150 ppm Amylene |
50-150 ppm Amylene |
50-150 ppm Amylene |
50 ppm Amylene |
| Water |
≤0.01% |
≤0.02% |
≤0.002% |
≤0.001% |
≤0.02% |
≤0.02% |
| |
|
|
≤0.005% (100mL) |
|
|
| Evaporation Residue |
≤0.0001% |
≤0.001% |
≤0.0003% |
≤0.0005% |
≤0.002% |
≤0.002% |
| Halogenated Residue |
≤10 ng/L (as Heptachlor epoxide) |
|
|
|
Passes Test |
Passes Test |
| Fluorescence 365 nm |
≤1.0 ppb |
|
| Titratable acid |
|
≤0.001% free acid (as HCL) |
|
|
≤0.0003 meq/g |
≤0.0003 meq/g |
| Free Halogens |
|
Passes Test |
Passes Test |
| Packaging Options |
| |
CHROMASOLV
Plus |
CHROMASOLV
for HPLC |
Biotech |
Anhydrous |
ACS
Spectro-
photometric |
ACS |
| Glass Bottle |
650463 |
34856 |
|
|
154792 |
D65100 |
| PVC Coated Bottle |
|
439223 |
|
|
|
320269 |
| Sure/Seal |
|
|
494453 |
270997 |
|
|
| Aluminum Bottle |
|
676853 |
| Pure-Pac |
|
34856 |
|
270997 |
|
|
| Carbon Steel |
|
D65100 |
| Drum |
|
439223 |
|
|
|
D65100 |
For a complete list of all available unit sizes and product specifications,
simply click on the product numbers above!