|
Dimethyl Sulfoxide Physical Properties |
 |
Dimethyl sulfoxide |
 |
 |
| |
|
| Sigma-Aldrich
Dimethyl sulfoxide
|
| Synonym |
Methyl sulfoxide |
Beilstein Registry |
506008 |
| Molecular Formula |
(CH3)2SO |
EG/EC Number |
2006643 |
| Molecular Weight |
78.13 |
MDL Number |
MFCD00002089 |
| CAS Number |
67-68-5 |
Vapor Pressure |
0.42
mm Hg (20°C) |
| Density |
1.100 g/mL at 25ēC |
Dielectric Constant |
47.24 at 20ēC |
| Boiling Point |
189ēC |
Dipole Moment |
3.960 |
| Melting Point |
16-19 ēC |
UV cutoff |
265 nm |
| Flash Point |
85.0°C |
Refractive Index |
1.479 at 20ēC |
| Viscosity |
1.990 at 25°C |
Vapor Density |
2.7 (vs. air) |
Dimethyl sulfoxide Specification Comparison Table