Sigma-Aldrich Dimethyl sulfoxide Specifications Comparison
|
| |
CHROMASOLV
Plus |
Biotech |
Anhydrous |
ACS
Spectro-
photometric |
ACS |
ReagentPlus |
| Purity |
≥99.7% |
99.98% |
≥99.9% |
≥99.9% |
≥99.9% |
≥99.6% |
| UV Absorption |
| λ: 270 nm |
≤0.70 |
≤1.00 |
|
≤0.40 |
|
|
| λ: 280 nm |
≤0.30 |
≤0.30 |
|
| λ: 290 nm |
|
|
|
≤0.18 |
|
|
| λ: 295 nm |
|
≤0.10 |
|
| λ: 300 nm |
≤0.10 |
|
| λ: 310 nm |
|
|
|
≤0.06 |
|
|
| λ: 330 nm |
|
|
|
≤0.02 |
|
|
| λ: 350 nm |
≤0.01 |
≤0.01 |
|
≤0.01 |
|
|
| λ: 400 nm |
|
≤0.01 |
|
≤0.01 |
|
|
| Total Impurities |
| Water |
≤0.2% |
≤0.005% |
≤0.005% |
≤0.1% |
≤0.1% |
≤0.1% |
| Evaporation Residue |
≤0.002% |
≤0.0005% |
≤0.0005% |
≤0.01% |
≤0.01% |
|
| Titratable acid |
|
|
|
≤0.001 meq/g |
≤0.001 meq/g |
|
| Packaging Options |
| |
CHROMASOLV
Plus |
Biotech |
Anhydrous |
ACS
Spectro-
photometric |
ACS |
ReagentPlus |
| Glass Bottle |
34869 |
|
|
154938 |
472301 |
471267 |
| Sure/Seal |
|
494429 |
276855 |
|
|
|
| VerSA-Flow |
|
472301 |
471267 |
| NOWPAK |
34869 |
|
| Pure-Pac |
|
494429 |
276855 |
|
|
|
| Drum |
34869 |
|
|
|
472301 |
|
For a complete list of all available unit sizes and product specifications,
simply click on the product numbers above!