Sigma-Aldrich
Research Essentials
Biological Buffers
Hematology and Histology
Solvents
2-Propanol Center
Acetone Center
Acetonitrile Center
Anhydrous BioTech
Bottles and Cans
CHROMASOLV
Chloroform Center
Dichloromethane Center
Dimethyl Sulfoxide Center
Ethyl Acetate Center
General Reagents
ACS Grade
Reagent Grade
Reagent Plus Grade
Hexane Center
High-Purity
Methanol Center
Semi Bulk
Tetrahydrofuran Center
Toluene Center
Water Center
Key Resources
Virtual Lab
Biochemicals
Chemicals
Key Resources
 General Reagents
Solvents
A.C.S. Solvents
Histological Solvents
Reagent Grade Solvents
A.C.S. Solvents
Complete list of ACS grade solvents
Sigma-Aldrich A.C.S. solvents meet or exceed the specifications set by the American Chemical Society. For those applications or general procedures requiring stringent quality specifications.
Sigma-Aldrich A.C.S. solvents are available in a wide variety of containers options including: Amber Glass Bottle, PVC Coated Glass Bottle, Safety Can, Carbon Steel Flex-Spout Can , Carbon Steel Can with NPT Threads, VerSA-Flow Container , Closed Head Steel Drum or Composite Drum.
Back to Top
Histological Solvents
Complete list of Histological grade solvents
Sigma-Aldrich provides a number of histological grade solvents commonly used for tissue sampling and processing. Precise control of the manufacturing process ensures unsurpassed lot-to-lot consistency.
We offer our Histological solvents in Amber Glass Bottle, Carbon Steel Flex-Spout Can, and Carbon Steel Can with NPT Threads.
Back to Top
Reagent Grade Solvents
Complete list of Reagent & ReagentPlus® solvents
We offer a wide variety of general applications solvents to meet your needs. ReagentPlus solvents have a guaranteed purity of ≥98.5% when higher purities are required such as specialty organic synthesis, preparative scale liquid chromatography or glass washing.
For general laboratory use, try our Reagent grade solvents; ≥95% purity.
Both of our Reagent and ReagentPlus solvents are available are available in a wide variety of containers options including: Amber Glass Bottle, PVC Coated Glass Bottle, Safety Can, Carbon Steel Flex-Spout Can, Carbon Steel Can with NPT Threads, VerSA-Flow Container, Closed Head Steel Drum or Composite Drum.
Back to Top