Sigma-Aldrich Hexane Specifications Comparison
|
| |
CHROMASOLV
Plus |
CHROMASOLV
for HPLC |
Anhydrous |
Spectro-
photometric |
ReagentPlus |
Reagent |
| Purity |
≥95% |
≥95% |
95% |
≥95% |
≥99% |
≥95% |
| UV Absorption |
| λ: 195 nm |
≤1.00 |
≤1.00 |
|
≤1.00 |
|
|
| λ: 200 nm |
≤0.50 |
|
| λ: 210 nm |
≤0.20 |
≤0.30 |
|
≤0.30 |
|
|
| λ: 220 nm |
≤0.05 |
≤0.10 |
|
≤0.10 |
|
|
| λ: 230 nm |
|
≤0.05 |
|
≤0.05 |
|
|
| λ: 240 nm |
|
≤0.02 |
|
≤0.02 |
|
|
| λ: 250 nm |
≤0.01 |
≤0.01 |
|
≤0.01 |
|
|
| λ: 280 nm |
≤0.005 |
|
| λ: 400 nm |
≤0.005 |
≤0.01 |
|
≤0.01 |
|
|
| Total Impurities |
| Water |
≤0.01% |
≤0.02% |
≤0.001% |
≤0.02% |
|
|
| |
|
≤0.005% (100mL) |
|
|
|
| Evaporation Residue |
≤0.0001% |
≤0.0005% |
≤0.0003% |
≤0.0005% |
|
|
| Halogenated Residue |
≤10 ng/L (as Heptachlor epoxide) |
|
| Fluorescence 365 nm |
≤1.0 ppb |
|
| Methylcyclopentane |
|
≤5% |
| Packaging Options |
| |
CHROMASOLV
Plus |
CHROMASOLV
for HPLC |
Anhydrous |
Spectro-
photometric |
ReagentPlus |
Reagent |
| Glass Bottle |
650552 |
270504 |
|
248878 |
139386 |
208752 |
| PVC Coated Bottle |
675393 |
439177 |
|
| Sure/Seal |
|
|
296090 |
|
|
|
| VerSA-Flow |
|
270504 |
|
| NOWPAK |
|
270504 |
|
| Carbon Steel |
|
208752 |
For a complete list of all available unit sizes and product specifications,
simply click on the product numbers above!