|
Hexane, Mixture of Isomers, Specification Comparison |
 |
Hexane, mixture of isomers |
 |
 |
| |
|
Printer friendly PDF version (120 kb)
Sigma-Aldrich Hexane Specifications Comparison
Mixture of isomers |
| |
CHROMASOLV
Plus |
CHROMASOLV
for HPLC |
Anhydrous |
ACS
Spectro-
photometric |
ACS |
| Purity |
≥98.5% |
≥98.5% |
≥99% |
≥98.5% |
≥98.5% |
| UV Absorption |
| λ: 195 nm |
≤1.00 |
≤1.00 |
|
≤1.00 |
|
| λ: 210 nm |
|
≤0.30 |
|
≤0.30 |
|
| λ: 220 nm |
|
≤0.10 |
|
≤0.10 |
|
| λ: 225 nm |
≤0.05 |
|
| λ: 230 nm |
|
≤0.05 |
|
≤0.05 |
|
| λ: 240 nm |
|
≤0.02 |
|
≤0.02 |
|
| λ: 250 nm |
≤0.01 |
≤0.01 |
|
|
|
| λ: 260 nm |
|
|
|
≤0.01 |
|
| λ: 275 nm |
≤0.005 |
|
| λ: 300 nm |
≤0.005 |
|
| λ: 400 nm |
|
≤0.01 |
|
≤0.01 |
|
| Total Impurities |
| Water |
≤0.01% |
≤0.02% |
≤0.001% |
≤0.02% |
|
| |
|
≤0.005% (100mL) |
|
|
| Evaporation Residue |
≤0.0001% |
≤0.0005% |
≤0.0005% |
≤0.0005% |
≤0.001% |
| Halogenated Residue |
≤10 ng/L (as Heptachlor epoxide) |
|
| Fluorescence 365 nm |
≤1.0 ppb |
|
| Titratable acid |
|
0.0003 meq/g |
|
0.0003 meq/g |
0.0003 meq/g |
| Sulfur compounds |
|
≤0.005% |
|
≤0.005% |
≤0.005% |
| Thiophene |
|
Passes Test |
|
Passes Test |
Passes Test |
| Packaging Options |
| |
CHROMASOLV
Plus |
CHROMASOLV
for HPLC |
Anhydrous |
ACS
Spectro-
photometric |
ACS |
| Glass Bottle |
650544 |
293253 |
|
156175 |
178918 |
| PVC Coated Bottle |
650420 |
439185 |
|
|
320315 |
| Sure/Seal |
|
|
227064 |
|
|
| Aluminum Bottle |
|
676799 |
| NOWPAK |
|
439185 |
|
|
|
| Pure-Pac |
|
293253 |
227064 |
|
|
| Carbon Steel |
|
178918 |
| Drum |
|
439185 |
|
|
178918 |
For a complete list of all available unit sizes and product specifications, simply click on the product numbers above!