|
Hexane Physical Properties |
 |
Hexane |
 |
 |
| |
|
| Sigma-Aldrich
Hexane |
| Synonym |
n-Hexane |
Beilstein Registry |
1730733 |
| Molecular Formula |
CH3(CH2)4CH3 |
EG/EC Number |
2037776 |
| Molecular Weight |
86.18 |
MDL Number |
MFCD00009520 |
| CAS Number |
110-54-3 |
Vapor Pressure |
132 mm Hg (20°C) |
| Density |
0.659 g/mL at 25ēC |
Dielectric Constant |
1.89 at 20ºC |
| Boiling Point |
69ēC |
UV cutoff |
210 nm |
| Melting Point |
-95ēC |
Refractive Index |
1.375 at 20ºC |
| Flash Point |
-23.3ēC |
Vapor Density |
3 (vs. air) |
| Viscosity |
0.300
at 25ēC |
Risk Statements |
11-38-48/20-51/53-62-65-67 |
| RIDADR |
UN 1208 3/PG 2 |
Safety Statements |
9-16-29-33-36/37-61-62 |
| Hazard Codes |
F,Xn,N |
 |
| Sigma-Aldrich
Hexane, mixture of isomers |
| Synonym |
Hexanes |
Beilstein Registry |
1730733 |
| Molecular Formula |
C6H14 |
EG/EC Number |
2035234 |
| Molecular Weight |
86.18 |
MDL Number |
MFCD00009520 |
| Density |
0.672
g/mL at 25ēC |
Dielectric Constant |
1.89 at 20ºC |
| Boiling Point |
68-70ēC |
UV cutoff |
195 nm |
| Melting Point |
-95ēC |
Refractive Index |
1.379 at 20ºC |
| Flash Point |
-22.7ēC |
Vapor Density |
3 (vs. air) |
| Viscosity |
0.286 at 25ēC |
Risk Statements |
11-38-48/20-51/53-62-65-67 |
| RIDADR |
UN 1208 3/PG 2 |
Safety Statements |
36/37-61-62 |
| Hazard Codes |
F,Xn,N |