Order Center
Research Essentials
Biological Buffers
Hematology and Histology
Solvents
2-Propanol Center
Acetone Center
Acetonitrile Center
Anhydrous BioTech
Bottles and Cans
CHROMASOLV
Chloroform Center
Dichloromethane Center
Dimethyl Sulfoxide Center
Ethyl Acetate Center
General Reagents
Hexane Center
High-Purity
Methanol Center
Semi Bulk
Tetrahydrofuran Center
Toluene Center
Water Center
Key Resources
Application Portal
FAQs
Me THF
Nomograph
Peroxide Formation
Production Capabilities
Purity Grades
Solvent Selector
Solvent Stabilizers
Virtual Lab
Biochemicals
Chemicals
Key Resources
 2-Methyltetrahydrofuran
An alternative to Tetrahydrofuran and Dichloromethane
2-Methyltetrahydrofuran (2-MeTHF) is a versatile, more environmentally friendly solvent derived from a variety of agricultural byproducts.
Features & Benefits
  • Quickly produces clean water phase splits (density of 0.86 g/mL at 25°C) without forming emulsions unlike THF or DCM
  • More easily dried than THF or DCM
  • Very stable even in highly acidic environments
  • Non-reactive, aprotic polar solvent
  • Forms an effective azeotrope with water
  • Higher boiling point (78-80°C)
      Lower solvent loss during reflux
      Allows rapid formation of Grignard reactants
  • Suitable replacement in most organometallic reactions where THF is used including Suzuki Coupling reactions
  Boiling Point Density g/mL at 25°C Flash Point Viscosity at 25°C Dielectric Constant at 25°C
2-Methyltetrahydrofuran 78-80°C 0.860 -12.0 0.600 6.97
Tetrahydrofuran 65-67°C 0.889 -17.2 0.456 7.52
Dichloromethane 39.8-40°C 1.325 none 0.413 8.93
2-Methyltetrahydrofuran
Products
Product #  Product Name Add to Cart
414247 anhydrous, ≥99.0%, contains 250ppm BHT as stabilizer
673277 anhydrous, ≥99%, Inhibitor-free
155810 ReagentPlus®, ≥98.5%, contains 150-400ppm BHT as stabilizer