Sigma-Aldrich
Research Essentials
Biological Buffers
Hematology and Histology
Solvents
2-Propanol Center
Acetone Center
Acetonitrile Center
Anhydrous BioTech
Bottles and Cans
CHROMASOLV
Chloroform Center
Dichloromethane Center
Dimethyl Sulfoxide Center
Ethyl Acetate Center
General Reagents
Hexane Center
High-Purity
Methanol Center
Semi Bulk
Tetrahydrofuran Center
Toluene Center
Water Center
Key Resources
Application Portal
FAQs
Me THF
Nomograph
Peroxide Formation
Production Capabilities
Purity Grades
Solvent Selector
Solvent Stabilizers
Virtual Lab
Biochemicals
Chemicals
Key Resources
 Peroxide–Forming Solvents
Solvents
A significant number of laboratory solvents can undergo autoxidation under normal storage conditions to form unstable and potentially dangerous peroxide by-products. This process is catalyzed by light and heat and occurs when susceptible materials are exposed to atmospheric oxygen. Molecular structure is the primary factor relating to a material’s potential for hazardous peroxide formation.
Most overviews of potential peroxide-forming chemicals classify various materials into three categories, on the basis of peroxide formation susceptibility, each with general handling and use guidelines.1 The two categories relevant to solvents are defined as solvents that pose a peroxide related safety risk with (Group B) or without (Group A) pre-concentration. Shelf life and storage information provided in Table I refers to time after opening the container and pertains to materials stored in opaque containers under inert atmospheric conditions.
Potential Peroxide-Forming Solvents*
Group A:  Chemicals that form explosive levels of peroxides without concentration
Severe peroxide hazard after prolonged storage, especially after exposure to air.
Test for peroxide formation before using or discard after 3 months.
  • Isopropyl ether
Group B:  Peroxide hazards on concentration
Test for peroxide formation before distillation or evaporation. Test for peroxide formation or discard after 1 year.
  • Acetal
  • Acetaldehyde
  • Benzyl Alcohol
  • 2-Butanol Dioxanes
  • Chlorofluoroethylene
  • Cumene(isopropylbenzene)
  • Cyclohexene
  • 2-Cyclohexen-1-ol
  • Cyclopentene
  • Decahydronapthalene(decalin)
  • Diacetylene(butadiyne)
  • Dicyclopentadiene
  • Diglyme
  • Diethyl ether
  • Ethylene glycol ether acetates
  • Furan
  • 4-Heptanol
  • 2-Hexanol
  • Methyl Acetylene
  • 3-Methyl-1-butanol
  • Methyl-isobutyl ketone
  • 4-Methyl-2-pentanol
  • 2-Pentanol
  • 4-Penten-1-ol
  • 1-Phenylethanol
  • Tetrahydrofuran
  • Tetrahydronapthalene
  • Vinyl Ethers
  • Sec. Alcohols
* Materials other than those listed may form peroxides. Applies only to pure materials.
Use and Storage of Peroxide-Forming Solvents
Common laboratory solvents found in Table I are generally Group B category and include diethyl ether, tetrahydrofuran, cyclohexene, glycol ethers, decalin and 2-propanol. Organic peroxides formed in these materials are significantly less volatile than the bulk solvent and, as a result, are susceptible to dangerous levels of peroxide enrichment via techniques such as evaporative concentration or distillation. In fact, most Group B solvents are sufficiently volatile that multiple openings of a container can be a sufficient source of peroxide concentration enrichment. The list below is a brief summary of key aspects of peroxide-forming solvent handling. For excellent review articles, the reader is referred to the work of Kelly2 or Clark3.
General Considerations for Peroxidizeable Solvents:
  • All peroxide-forming solvents should be checked for the presence of any peroxides prior to distillation or evaporation.
  • Solvents containing low levels of free radical scavengers such as BHT should be used whenever the presence of the stabilizing species does not interfere with intended application.
  • Uninhibited materials should be stored with care and frequently checked for peroxide formation.
  • Peroxide-forming solvents should be purchased in limited quantities and older material in inventory should be preferentially selected for use.
  • Materials should be stored away from light and heat with tightly secured caps and labeled with dates of receipt and opening.
  • Periodic testing to detect peroxides should be performed and recorded on previously opened material.
Peroxide Detection
A variety of methods are available to test for the presence of peroxides in organic solvents with the two most common tests described below. At present, 100 ppm is widely used as a general control point with respect to minimum hazardous peroxide concentration in a solvent, however, this value lacks scientific validation and is likely too liberal or conservative depending on the solvent in question and intended application.2 Do not test any peroxide-forming chemicals if there are visible crystals, visible precipitate or an oily viscous layer present in the material since these are visual indicators of dangerous peroxide levels.
Quantofix® Peroxide Test Strips (Sigma-Aldrich Part # 37206)
In the presence of hydrogen peroxide the test paper turns blue. Quantofix® Peroxide test sticks can also be used for the determination of peracetic acid and other organic and inorganic hydroperoxides. To test for hydroperoxides in organic solvents, the test zone is wetted with one drop of water after evaporation of the solvent.
Interferences:  In the pH range of 2-9, the accuracy of the determination is independent of the pH of the test solution. Buffer strongly acidic solutions with sodium acetate and adjust alkaline solutions to about pH 5-7 with citric acid. Falsely positive results can only be caused by strong oxidizing agents.
Storage:  Avoid exposing the sticks to sunlight and moisture. Store unopened packs in refrigerator (+2 °C to +8 °C). Opened packs: store container in a cool and dry place.
Potassium Iodide Indicator
Add 0.5-1.0 ml of the sample solvent to an equal volume of glacial acetic acid containing about 0.1 g of sodium iodide or potassium iodide crystals. A yellow color indicates iodine formation via iodide oxidation by sample peroxide; a brown color indicates high concentration. A blank determination should be made particularly when color development is faint since iodide/acetic acid mixtures will, over time, turn a yellow - brown color due to air oxidation.
A more sensitive variation of the above method adds one drop of a saturated, aqueous starch solution to the sample solution. Starch and iodine combine to form a bright blue complex that is more easily visualized than the yellow color generated by iodine alone. Dark blue solution color would be indicative of high peroxide concentrations.
References
  1. Jackson, H.L., McCormack, W.B., Rondestvedt, C.S., Smeltz, K.C., and Viele, I.E. Control of Peroxidizable Compounds, J. Chem. Educ., 1970, 46 (3), A175.
  2. Kelly, R.J, Review of Safety Guidelines for Peroxidizable Organic Compounds, Chemical Health and Safety, 1996, 3 (5), 28-36.
  3. Clark, D.E., Peroxides and Peroxide - Forming Compounds, Chemical Health and Safety, 2001, 8 (5), 12-21.