Sigma-Aldrich
Research Essentials
Biological Buffers
Hematology and Histology
Solvents
2-Propanol Center
Acetone Center
Acetonitrile Center
Anhydrous BioTech
Bottles and Cans
CHROMASOLV
Chloroform Center
Dichloromethane Center
Dimethyl Sulfoxide Center
Ethyl Acetate Center
General Reagents
Hexane Center
High-Purity
Methanol Center
Semi Bulk
Tetrahydrofuran Center
Toluene Center
Water Center
Key Resources
Application Portal
FAQs
Me THF
Nomograph
Peroxide Formation
Production Capabilities
Purity Grades
Solvent Selector
Solvent Stabilizers
Virtual Lab
Biochemicals
Chemicals
Key Resources
 Pressure-Temperature Nomograph
Solvents
Sigma-Aldrich Pressure Temperature NomoGraph
Quickly and easily estimate boiling points at various pressures. The interactive Pressure-Temperature Nomograph simplifies calculations to improve the efficiency of your distillation or evaporation.
Launch NomoGraph
Physical Properties tab
Pressure Conversion Tab: Use the built-in Pressure Conversion Calculator which can convert between five units of pressure. Simply enter a numeric value or scientific notation and calculate.
Temperature Conversion Tab: Quickly calculate temperature conversions without leaving the Nomograph.
Physical Properties Tab: The Pressure-Temperature Nomograph provides physical data (650k PDF) for all your common solvents, including a list of relevant products.
Calculate a boiling point or pressure using the Antoine Equation: The Pressure-Temperature Nomograph is a graphical application of the Clausius-Clapeyron Equation, which assumes the heat of vaporization is a constant over a pressure range. Antoine´s Equation gets around this by using empirical data for each unique liquid under consideration.
Printable: Need a hard copy to take with you? Simply right-click and select 'print'.
Get Adobe Flash Player
This presentation requires the Flash plug-in for proper viewing. Download the player FREE.