Sigma-Aldrich
Research Essentials
Biological Buffers
Hematology and Histology
Solvents
2-Propanol Center
Acetone Center
Acetonitrile Center
Anhydrous BioTech
Bottles and Cans
CHROMASOLV
Chloroform Center
Dichloromethane Center
Dimethyl Sulfoxide Center
Ethyl Acetate Center
General Reagents
Hexane Center
High-Purity
Methanol Center
Semi Bulk
Cylinder Return
Drums
Flex Spout
NPT Threaded Cans
NowPak Containers
PurePac
VerSA-Flow
Tetrahydrofuran Center
Toluene Center
Water Center
Key Resources
Virtual Lab
Biochemicals
Chemicals
Key Resources
 Semi-Bulk
Solvents
Sigma-Aldrich offers a variety of packaging options for those applications when individual bottles are not practical. With larger volumes of solvents, safe and proper dispensing methods are essential.
Pure-Pac™ Returnable Containers
Carbon Steel Flex-Spout Can
Carbon Steel Can with NPT Threads
VerSA-Flow™
NOWPak® Containers
Drums
Packaging Specifications
Pure-Pac™ Returnable Containers
Products available in Pure-Pac containers
From Research to Production scale volumes, our Stainless Steel Pure-Pac fleet ensures your Sigma-Aldrich solvents maintain their high level of purity and dryness through a positive pressure, closed transfer design.
Cylinder Return Program
Returnable Container FAQs
Adaptability, environmental issues, increasing operating costs and the need to find a solution for the inefficiencies of the 4L glass bottles make the Pure-Pac an outstanding alternative to traditional solvent packaging. There are no empty bottles to rinse and dispose of and the larger batch size reduces the Quality Control burden.
Safety Features
The Pure-Pac containers will enhance operator safety when handling and dispensing large volumes of high purity solvents.
  • Rugged stainless steel construction reduces the likelihood of hazards associated with spills or bottle breakages
  • Dispensing under a positive pressure, closed system virtually eliminates exposure to hazardous solvent vapors
  • To avoid over-pressurization, each container has a built-in pressure relief for the utmost in safety
  • To monitor the container pressure, a easy to read pressure gauge is located atop of the Pure-Pac
  • FM approval (Pure-Pac I)
Technical Bulletin AL-201 (676 Kb PDF) and Technical Bulletin AL-236 (604 Kb PDF) are available to assist with:
  • Recommended Transfer Procedures
  • Storage
  • Related Equipment
  • Return Policy
Pure-Pac I
Pure-Pac I
  Specifications for the Sigma-Aldrich Pure-Pac I Containers
  18L Pure-Pac 200L Pure-Pac 400L Pure-Pac
Total capacity, (L) 20 230 438
Working capacity, (L) 18 200 400
UN Specification UN1A1 UN1A1 UN1A1
ASME Code Section VIII Div. 1 Section VIII Div. 1 Section VIII Div.
Maximum use pressure (psig) 15 15 15
Pressure relief (psig) 40 40 40
Maximum container pressure (psig) 100 50 50
Tare weight with
valves (kg)
17 80 105
Shipping cube
L x W x H (cm)
33.02x33.02x57.15 71.12x71.12x144.78 71.12x71.12x162.56
Fork truck/forklift access not included included included
Nitrogen inlet/vent
fitting
3/8" ferrule lock 3/8" ferrule lock 3/8" ferrule lock
Solvent outlet fitting 3/8" ferrule lock 1/2" ferrule lock 1/2" ferrule lock
Nitrogen flushing
port fitting
3/8" ferrule lock 1/2" ferrule lock 1/2" ferrule lock
Valve configuration for Pure-Pac I
Pure-Pac II
Pure-Pac II
Specifications for the Sigma-Aldrich Pure-Pac II Containers
  20L Pure-Pac II 50L Pure-Pac II 200L Pure-Pac II
Total capacity, (L) 22 55 220
Working capacity, (L) 20 50 200
UN Specification UN1A1 UN1A1 UN1A1
Maximum use pressure (psig) 5 5 5
Pressure relief (psig) 12 12 12
Maximum container pressure (psig) 40 40 40
Tare weight (kg) 7.3 14 45
Shipping cube
L x W x H (cm)
27.94x27.94x50.8 40.64x40.64x58.42 60.96x60.96x96.52
Nitrogen inlet/vent
fitting
1/4" FNPT 1/4" FNPT 1/4" FNPT
Solvent outlet fitting 1" MNPT 1" MNPT 1" MNPT
Configuration for Pure-Pac II
As these are highly specialized, product dedicated pressure rated containers, a deposit is required upon purchase. Each container is stamped with a unique serial number and a detailed individual history is maintained. Sigma-Aldrich has a complete maintenance program to ensure that each customer receives a deposit container in the best possible condition. Because the Pure-Pac container is highly specialized, we ask they be returned immediately after they have been emptied.
For more details regarding Pure-Pac containers, please feel free to contact us at labessentials@sial.com.
Back to Top
Carbon Steel Flex-Spout Can
Products available in Flex-Spout cans
Solid carbon steel design comes with a convenient built-in spout for easy of dispensing. Ideal for use with many of our ACS, Reagent and Histological Grade Solvents.
  • 20 Liter capacity
  • Polyethylene Flex-Spout
  • UN and DOT approved UN1A1/X1.4/250
  • 36.35cm H 27.94cm D
Dispensing Methods
Pouring - Comes with built-in one-piece self-venting polyethylene spout for easy pouring.
Pumping
Flex-Spout Pump (Z557803)
This Rieke pump is for dispensing solvents and non-corrosive liquids. The self-venting design eliminates a need for an air vent opening. Pump includes a plastic Flex-Spout mounting cap and a stabilizing metal strap for mounting to Sigma-Aldrich 20L Flex-Spout cans. Dispenses ~100mL per stroke.
Carbon Steel Pump (Z555363)
High quality, FM Approved pump with grounding wires and clamps makes for the safe dispensing of solvents and non-corrosive liquids. Pump includes a set of cap adapters for mounting to various cans including the Sigma-Aldrich 20L Flex-Spout can. Dispenses ~100mL per stroke.
Back to Top
Carbon Steel Can with NPT Threads
Products available in NPT threaded cans
Solid carbon steel design incorporates a 2" and 3/4" NPT thread for the dispensing of our ACS, Reagent and Histological Grade Solvents.
  • 20 Liter capacity
  • UN and DOT approved UN1A1/X1.4/250
  • 36.35cm H x 27.94cm D
Dispensing Methods
Pouring
Pump N' Pour Adapter (Z557757)
Threaded 2" NPT polypropylene adapter allows for the pouring of solvents directly from the container. Comes with o-ring cap for tight closure when not in use. See AL-222 for instructions on installation.
Pour Spout (Z511870)
Low-density polyethylene spout fits over the can’s 2" NPT opening.
Faucet
Brass faucet (Z160482) Self-closing, FM approved, fits 3/4" NPT thread
Self-venting PP faucet (Z168149) fits 3/4" NPT thread
Pumping
VerSA-Flow Tapper (Z529060)
Simple and convienent pumping action pressurizes container for continuous flow.
  • PTFE solvent pick-up tube and On/Off spigot
  • HDPE body and piston with PTFE wiper
Flex-Spout Pump (Z557803)
This Rieke pump is for dispensing solvents and non-corrosive liquids. The self-venting design eliminates a need for an air vent opening. Dispenses ~100mL per stroke. For tight seal, use in conjunction with Flex-Spout NPT Adapter (Z557811)
Carbon Steel Pump (Z555363)
High quality, FM approved pump with grounding wires and clamps makes for the safe dispensing of solvents and non-corrosive liquids. Carbon Steel Pump Adapter (Z562254) is required for use with Sigma-Aldrich 20L NPT can with 2 inch bung (FM approved). Dispenses ~100mL per stroke.
Pressure Transferring
VerSA-Flow Economy Pressure Delivery Cap (Z529028)
Solid PBT machined body with two tubing ports.
  • Flow rate ~1L/min. at 4psi.
  • Includes 10 ft x 1/4 in. O.D. FEP™ tubing
  • Use at <5psi max.
VerSA-Flow Low-Flow Pressure Delivery Cap (Z545732)
Two-piece, PBT cap permits its removal without disconnecting solvent and gas lines when changing containers.
  • Flow rate ~1L/min. at 4psi.
  • Four 1/4 in. NPT threaded holes with plugs permit the solvent lines, liquid-level indicator, vent line, or sparge/delivery tube assemblies
  • Built-in pressure-relief valve automatically vents in case of excessive pressure (5 psi max.).
  • Includes 10 ft x 1/4 in. O.D. FEP™ tubing
VerSA-Flow High-Flow Pressure Delivery Cap (Z545791)
Two-piece, PBT cap permits the removal of the cap without disconnecting solvent and gas lines when changing containers.
  • Flow rate: ~5 to 6 L/min @ 4 psi
  • Built-in pressure-relief valve vents automatically in case of excessive pressure (10 psi max.)
  • Includes 8 ft x 3/8 in. I.D. FEP
  • PTFE On/Off spigot
See AL-210 for all VerSA-Flow instructions, installation, and additional dispensing need options.
Accessories
Plug Wrench (Z219290)
Nonsparking bronze alloy. Fits most bung patterns.
Flexible bonding wire (Z197866)
Prevents sparking of static electricity generated by the transfer of flammable liquids. 3ft. L with 2 copper alligator clips.
Back to Top
VerSA-Flow™
VerSA-Flow™ Container
Products available in VerSA-Flow containers
This 18L HDPE liner with metal overpack can is used for many of our ACS, CHROMASOLV® for HPLC, Reagent grade and Mobile Phase Solvent Blends. Easily configured to be used with HPLC analysis.
  • Double containment design for safe shipping and storage
  • Completely recyclable
  • Convenient for holding waste solvents when emptied
  • UN and DOT approved UN1A2/X50/S
  • 2 in. buttress thread
  • 41.28cm H x 29.85cm D
  Dispensing Methods
Pumping
VerSA-Flow Tapper (Z529060)
Simple and convienent pumping action pressurizes container for continuous flow.
  • PTFE solvent pick-up tube and On/Off spigot
  • HDPE body and piston with PTFE wiper
  • Requires (Z529036) Buttress Adapter
Pressure Transferring
VerSA-Flow Economy Pressure Delivery Cap (Z529028)
Solid PBT machined body with two tubing ports.
  • Flow rate ~1L/min. at 4psi.
  • Includes 10 ft x 1/4 in. O.D. FEP™ tubing
  • Use at <5psi max.
  • Requires (Z529036) Buttress Adapter
VerSA-Flow Low-Flow Pressure Delivery Cap (Z543810)
Two-piece, PBT cap permits its removal without disconnecting solvent and gas lines when changing containers.
  • Flow rate ~1L/min. at 4psi.
  • Four 1/4 in. NPT threaded holes with plugs permit the solvent lines, liquid-level indicator, vent line, or sparge/delivery tube assemblies
  • Built-in pressure-relief valve automatically vents in case of excessive pressure (5 psi max.).
  • Includes 10 ft x 1/4 in. O.D. FEP™ tubing
Low-Flow Pressure Delivery Cap Accessories
See AL-210 for all VerSA-Flow instructions, installation, and additional dispensing need options.
HPLC Tube Fittings (Z543853)
Use compressions fittings to connect directly to your HPLC instrument’s 1/8 in. tubing.
Liquid Level Indicator (Z543780)
316 SS, color-coded indicator wire for visual determination of solvent level inside the container
Sparge/Solvent delivery assembly (Z543829)
2 micron SS filter, 1/8 in. o.d. PFA tubing
Single hole metal cover with locking ring (Z553190)
Allows for a solid, locked cover to be used on the VerSA-Flow container while dispensing is in process.
VerSA-Flow High-Flow Pressure Delivery Cap (Z553182)
Two-piece, PBT cap permits the removal of the cap without disconnecting solvent and gas lines when changing containers.
  • Flow rate: ~5 to 6 L/min @ 4 psi
  • Built-in pressure-relief valve vents automatically in case of excessive pressure (10 psi max.)
  • Includes 8 ft x 3/8 in. I.D. FEP
  • PTFE On/Off spigot
Back to Top
NOWPak® Containers
NOWPak Container Image
Products available in NOWPak® containers
Both the 10L and 20L sizes incorporate a pre-cleaned, inert PFA liner, which is virtually compatible with all high-purity solvents. The NOWPak containers feature sturdy High-Density Polyethylene over packs providing secondary solvent containment during shipping, storage and handling. They are ideal for our CHROMASOLV® Solvent line including the Mobile Phase Solvent Blends used in HPLC analysis.
  • Secondary solvent containment design
  • Completely recyclable
Bottle/Packaging Specifications
Capacity Bottle Dimension
(H x D) cm
Closure Case Pack Case Pack Dimension
(L x W x H) cm
UN/DOT Rating
10L 33.24 x 22.95 63 mm 2 x 10L 48.26 x 25.40 x 37.47 4G/X42.8/S
20L 41.43 x 30.48 63 mm N/A N/A 1H1/Y1.9/100
  Dispensing Methods
Pumping
Cap Connector for 1/4 in. Tubing ND-LD-00 (Z560073)
Provides sealed solvent dispensing when used with pump or vacuum dispenser.
  • 1/4 in. tube elbow fitting
  • Removable FEP dip tube
  • 40 micron stainless steel filter cup
  • PTFE, FEP, PFA wetted materials
Cap Connector for 1/8 in. Tubing ND-LD-14 (Z560081)
Ideal for use with HPCL instrumentation where solvent sparging is not required.
  • 1/8 in. tube elbow fitting
  • Removable FEP dip tube
  • 40 micron stainless steel filter cup
  • PTFE, FEP, PFA wetted materials
Pressure Transferring
Pressure Dispenser Connector ND-LD-18 (Z560103)
Designed for closed systems pressure transfers of solvents.
  • 1/4 in. tube elbow fitting
  • Pressure rated blue gas line
  • 10psi pressure relief valve
  • Removable FEP dip tube
  • 40 micron stainless steel filter cup
  • PTFE, FEP, PFA wetted materials
Back to Top
Drums
  Sigma-Aldrich solvents are packaged in both HPDE lined and carbon steel drums for a limited offering of our ReagentPlus, ACS, CHROMASOLV and CHROMASOLV Blends.
Products available in drums
If you are looking for additional solvents in drum quantities, please contact us at labessentials@sial.com.
Drum Specifications
  Drum Lining Dimension (HxD) cm Closure UN/DOT Rating
50L Composite HDPE 66.68 x 36.83 2" NPT UN6HA1/Y2.0/150
50L Closed Head Carbon steel 55.25 x 46.99 2" & 3/4" NPT UN1A1/X1.2/300
200L Composite HDPE 92.08 x 62.23 2" NPT UN6HA1/Y1.2/150
200L Closed Head Carbon Steel 88.27 x 58.42 2" & 3/4" NPT UN1A1/X1.5/300
Dispensing Methods
Pumping
Heavy-duty pump (Z200506)
High quality, 316 stainless steel construction with a PTFE diaphragm. Fits the 2" bung of both the 50L and 200L drum. Dispenses ~16oz of solvent per down stroke.
Pressure Transferring
VerSA-Flow Economy Pressure Delivery Cap (Z529028)
Solid PBT machined body with two tubing ports.
  • Flow rate ~1L/min. at 4psi.
  • Includes 10 ft x 1/4 in. O.D. FEP™ tubing
  • Use at <5psi max.
VerSA-Flow Low-Flow Pressure Delivery Cap (Z545732)
Two-piece, PBT cap permits its removal without disconnecting solvent and gas lines when changing containers.
  • Flow rate ~1L/min. at 4psi.
  • Four 1/4 in. NPT threaded holes with plugs permit the solvent lines, liquid-level indicator, vent line, or sparge/delivery tube assemblies
  • Built-in pressure-relief valve automatically vents in case of excessive pressure (5 psi max.).
  • Includes 10 ft x 1/4 in. O.D. FEP™ tubing
VerSA-Flow High-Flow Pressure Delivery Cap (Z545791)
Two-piece, PBT cap permits the removal of the cap without disconnecting solvent and gas lines when changing containers.
  • Flow rate: ~5 to 6 L/min @ 4 psi
  • Built-in pressure-relief valve vents automatically in case of excessive pressure (10 psi max.)
  • Includes 8 ft x 3/8 in. I.D. FEP
  • PTFE On/Off spigot
See AL-210 for all VerSA-Flow instructions, installation, and additional dispensing need options.
Accessories
Drum level gauge (Z202096)
For gauging contents of drums draining in a horizontal position. Fits 3/4" NPT bung opening. Constructed of heavy gauge glass mounted in plated steel body with brass fitting. Buna-N® seals. Requires a 3/4" NPT safety drum faucet.
Brass faucet (Z160482)
Self-closing, FM approved, fits 3/4" NPT thread
Dual-action drum vent (Z197750)
Automatic pressure relief valve opens at 5 psi internal pressure. Vacuum relief is automatic and permits 4 gpm flow through a standard 3/4 in. faucet. HDPE with steel flange.
Back to Top