|
Tetrahydrofuran Physical Properties |
 |
Tetrahydrofuran |
 |
 |
| |
|
| Sigma-Aldrich
Tetrahydrofuran |
| Synonym |
THF |
Beilstein Registry |
102391 |
| Molecular Formula |
C4H8O |
EG/EC Number |
2037268 |
| Molecular Weight |
72.11 |
MDL Number |
MFCD00005356 |
| CAS Number |
109-99-9 |
Vapor Pressure |
143
mm Hg (20°C) |
| Density |
0.
889
g/mL at 25ēC |
Dielectric Constant |
7.52
at 22ēC |
| Boiling Point |
65-67ēC |
Dipole Moment |
1.750 |
| Melting Point |
-108ēC |
UV cutoff |
220
nm |
| Flash Point |
-17.2ēC |
Refractive Index |
1.407
at 20ēC |
| Viscosity |
0.456
at 25ēC |
Vapor Density |
2.5 (vs. air) |
| RIDADR |
UN 2056 3/PG 2 |
Risk Statements |
11-19-36/37 |
| Hazard Codes |
F,Xi |
Safety Statements |
16-29-33 |
 |
Tetrahydrofuran Specification Comparison Table