Order Center
Research Essentials
Biological Buffers
Hematology and Histology
Solvents
2-Propanol Center
Acetone Center
Acetonitrile Center
Anhydrous BioTech
Bottles and Cans
CHROMASOLV
Chloroform Center
Dichloromethane Center
Dimethyl Sulfoxide Center
Ethyl Acetate Center
General Reagents
Hexane Center
High-Purity
Methanol Center
Semi Bulk
Tetrahydrofuran Center
Miscibility / Immiscibility
Physical Properties
Specification Comparison
Toluene Center
Water Center
Key Resources
Virtual Lab
Biochemicals
Chemicals
Key Resources
 Tetrahydrofuran Specification Comparison
Tetrahydrofuran
Tetrahydrofuran (THF) Specification Comparison Printer friendly PDF version (120 kb)
Sigma-Aldrich Tetrahydrofuran Specifications Comparison
  CHROMASOLV
Plus
Biotech Anhydrous Anhydrous Spectro-
photometric
ACS ReagentPlus
Purity ≥99.9% ≥99.9% ≥99.9% ≥99.9% ≥99.5% ≥99% ≥99%
UV Absorption
λ: 212 nm   ≤1.00     ≤1.00    
λ: 245 nm ≤0.30      
λ: 250 nm   ≤0.18     ≤0.18    
λ: 275 nm ≤0.05            
λ: 300 nm   ≤0.02     ≤0.02    
λ: 315 nm ≤0.004            
λ: 350 nm   ≤0.005     ≤0.005    
λ: 400 nm   ≤0.005     ≤0.005    
Total Impurities
Inhibitor Inhibitor-free Inhibitor-free Inhibitor-free 250 ppm BHT Inhibitor-free 250 ppm BHT 250 ppm BHT
Water ≤0.05% ≤0.005% ≤0.002% ≤0.002% ≤0.02% ≤0.05%  
    ≤0.005% (100mL) ≤0.005% (100mL)      
Evaporation Residue ≤0.001%   ≤0.0005%   ≤0.0005% ≤0.03%  
Titratable acid ≤0.002% free acid (as CH3COOH)            
Peroxide =0.05% peroxide (as H2O2) ≤0.005% peroxide (as H2O2) ≤0.002% peroxide (as H2O2)   ≤0.015% peroxide (as H2O2) ≤0.015% peroxide (as H2O2)  
Packaging Options
  CHROMASOLV
Plus
Biotech Anhydrous Anhydrous Spectro-
photometric
ACS ReagentPlus
Glass Bottle 34865       242888 360589 178810
PVC Coated Bottle 439215         437638  
Sure/Seal   494461 401757 186562      
Aluminum Bottle         676764  
NOWPAK 34865            
Pure-Pac 34865 494461 401757 186562      
Carbon Steel           360589 178810
Drum           360589  
For a complete list of all available unit sizes and product specifications,
simply click on the product numbers above!