|
Toluene Physical Properties |
 |
Toluene |
 |
 |
| |
|
| Sigma-Aldrich
Toluene |
| Synonym |
Methylbenzene |
Beilstein Registry |
635760 |
| Molecular Formula |
C6H5CH3 |
EG/EC Number |
2036259 |
| Molecular Weight |
92.14 |
MDL Number |
MFCD00008512 |
| CAS Number |
108-88-3 |
Vapor Pressure |
22
mm Hg (20°C) |
| Density |
0.
865
g/mL at 25ēC |
Dielectric Constant |
2.38
at 23ēC |
| Boiling Point |
110-111ēC |
Dipole Moment |
0.375 |
| Melting Point |
-93ēC |
UV cutoff |
286
nm |
| Flash Point |
4.4ēC |
Refractive Index |
1.496
at 20ēC |
| Viscosity |
0.560
at 25ēC |
Vapor Density |
3.2 (vs. air) |
| RIDADR |
UN 1294 3/PG 2 |
Risk Statements |
11-38-48/20-63-65-67 |
| Hazard Codes |
F,Xn |
Safety Statements |
36/37-46-62 |
 |
Toluene Specification Comparison Table