Sigma-Aldrich Toluene Specifications Comparison
|
| |
CHROMASOLV
Plus |
CHROMASOLV
for HPLC |
Anhydrous |
ACS
Spectro-
photometric |
ACS |
ReagentPlus |
| Purity |
≥99.9% |
99.9% |
99.8% |
≥99.5% |
≥99.5% |
99% |
| UV Absorption |
| λ: 284 nm |
≤1.00 |
|
| λ: 286 nm |
|
|
|
≤1.00 |
|
|
| λ: 288 nm |
|
|
|
≤0.50 |
|
|
| λ: 290 nm |
|
≤0.30 |
|
| λ: 293 nm |
|
|
|
≤0.20 |
|
|
| λ: 300 nm |
≤0.10 |
|
|
≤0.10 |
|
|
| λ: 310 nm |
|
≤0.05 |
|
≤0.05 |
|
|
| λ: 325 nm |
≤0.02 |
|
| λ: 335 nm |
|
|
|
≤0.02 |
|
|
| λ: 350 nm |
≤0.005 |
≤0.004 |
|
≤0.01 |
|
|
| λ: 400 nm |
≤0.005 |
|
|
≤0.01 |
|
|
| Total Impurities |
| Water |
≤0.02% |
≤0.02% |
≤0.001% |
≤0.03% |
≤0.03% |
|
| |
|
≤0.005% (100mL) |
|
|
|
| Evaporation Residue |
≤0.0001% |
≤0.0005% |
≤0.0005% |
≤0.0004% |
≤0.001% |
|
| Halogenated Residue |
≤10 ng/L (as Heptachlor epoxide) |
|
| Titratable acid |
|
≤0.001% free acid (as HCl) |
|
| Sulfur compounds |
|
|
|
≤0.003% |
≤0.003% |
|
| Thiophene |
|
≤0.0005% |
|
| Packaging Options |
| |
CHROMASOLV
Plus |
CHROMASOLV
for HPLC |
Anhydrous |
ACS
Spectro-
photometric |
ACS |
ReagentPlus |
| Glass Bottle |
650579 |
34866 |
|
155004 |
179418 |
179965 |
| PVC Coated Bottle |
|
648566 |
|
|
320552 |
|
| Sure/Seal |
|
|
244511 |
|
|
|
| Aluminum Bottle |
|
676756 |
|
| NOWPAK |
|
34866 |
|
| Pure-Pac |
|
34866 |
244511 |
|
|
|
| Carbon Steel |
|
179418 |
179965 |
For a complete list of all available unit sizes and product specifications,
simply click on the product numbers above!