Water Physical Properties |
 |
Water |
 |
 |
| |
|
| Sigma-Aldrich
Water |
| Molecular Formula |
H2O |
Beilstein Registry |
2050024 |
| Molecular Weight |
18.02 |
EG/EC Number |
2317912 |
| CAS Number |
7732-18-5 |
MDL Number |
MFCD00011332 |
| Density |
1.000 g/mL at 3.98 °C |
Dielectric Constant |
80.10 at 20°C |
| Boiling Point |
100°C |
Dipole Moment |
1.82 |
| Melting Point |
0.0°C |
UV cutoff |
191 nm |
| Viscosity |
1.000 at 20°C |
Refractive Index |
1.340 at 20°C |
Water Specification Comparison Table