Order Center
The Americas
Argentina
Brazil
Canada
About Us
Customer Service
New Lab Form
New Lab Startup Program
Online Help & Hints
Online Services
Ordering
Sigma Genosys Canada
Terms & Conditions
Canada (Francais)
Mexico
US Export
United States
Promotion Code C04
Canada
July 31,2008
Canada
Custom qPCR Probes
For a limited time, receive discounted pricing
Features of our Custom Oligo Service
  • Dual-labeled probes are shipped in amber tubes to ensure dye integrity.
  • Every dual-labeled probe is purified by reverse-phase HPLC.
  • All oligos are subject to 100% QC by MALDI-TOF mass spectrometry and/or CE.
  • Standard delivery is 3–5 working days for FAM, HEX, TET, Fluorescein combinations.
  • An additional vial label is included for every oligo.
DUAL-LABELED PROBES 1 OD($) 3 OD($) 5 OD($) 10 OD($)
5’-end 6-FAM / 3’-end BHQ-1 or TAMRA 73.15 134.75 227.15 365.75
5’-end Cy3/ 3’-end BHQ-2 173.25 250.25 365.75 539.00
5’-end Cy5.5/ 3’-end BHQ-2 173.25 250.25 365.75 539.00
5’-end Cy5/ 3’-end BHQ-2 173.25 250.25 365.75 539.00
5’-end Fluorescein / 3’-end BHQ-1or TAMRA 73.15 134.75 227.15 365.75
5’-end HEX / 3’-end BHQ-1 or TAMRA 73.15 134.75 227.15 365.75
5’-end JOE/ 3’-end BHQ-1 or TAMRA 304.15 458.15 689.15 1035.65
5’-end ROX / 3’-end BHQ-2 304.15 458.15 689.15 1035.65
5’-end TET / 3’-end BHQ-1 or TAMRA 73.15 134.75 227.15 365.75
5’-end TAMRA / 3’-end BHQ-2 173.25 250.25 365.75 539.00
5’-end Texas Red / 3’-end BHQ-2 304.15 458.50 689.15 1035.65
Must reference promo C04 when ordering.Valid in Canada only. Expires 7/31/08
Primers can be ordered at a discounted price of $0.24/base on the 3 OD yield or$0.27/base on the 5 OD yield. Oligo length must be less than or equal to 50 bases to qualify, and primers must be ordered with duallabeled probes. Price includes 5’ – end Reporter, 3’ – end Quencher, HPLC purification, and base charges.Price applies to probes 15–40 bases in length.
Sigma offers a broad range of options for your probe-based applications including:
Dual-labeled fluorogenic probes
  • Molecular beacons
  • LNAฎ containing probes
  • Light Cyclerฎ Probes
  • Scorpions™ Probes
Ordering: sigma.com/oligos
customdna@sial.com
Fax: (866) 677-0377
or Call (866) 677-0277