Molecular Biology
Automated Sequencing
Cloning and Expression
DNA and RNA Purification
Gene Expression Analysis
Molecular Biology Reagents
Nucleic Acid Detection
Nucleic Acid Electrophoresis
PCR
Product Lines
New PCR Products
Key Resources
Whole Genome Amplification
New Products
Neuroscience
Nutrition Research
Obesity Research
Peptides and Proteins
Plant Biotechnology
Proteomics and Protein Expr.
Stable Isotopes
Stem Cell Biology
Your Favorite Gene - Search
Life Science Innovations
PathFinder
 Hot Start PCR
PCR
Hot Start PCR techniques focus on the inhibition of polymerase activity during reaction preparation. By limiting polymerase activity prior to PCR cycling, non-specific amplification is reduced and target yield is increased. Common methods for Hot Start PCR include chemical modifications, wax-barrier methods, and inhibition by a taq-directed antibody.
Sigma's JumpStart Taq polymerase is an antibody inactivated hot start enzyme designed to minimize non-specific amplification while increasing target yield. Unlike other Hot Start methods (i.e. chemical inactivation), JumpStart Taq polymerase does not require a pre-incubation step prior to cycling because polymerase activity is fully restored during the first denaturation cycle of the PCR reaction.
Sigma JumpStart Taq Enzymes & ReadyMixes:
  • Greater Specificity & Increased Target Yield. JumpStart Taq antibody prevents non-specific amplification while increasing target yield.
  • Enhanced Sensitivity. JumpStart Taq DNA Polymerases provide superior amplification regardless of template concentration.
  • Flexibility to Meet Reaction Needs. Formulated for a variety of PCR needs including the ease of direct load with REDTaq reagents or the convenience of our ReadyMixes that only require the addition of primers, template, and water.
  • Convenience. Reduce set-up time, eliminate contamination concerns, and avoid long reactivation steps with JumpStart Reagents & Readymixes.
Product Table Technical Data Special Offers
Product Name Technical Bulletin Product # 
JumpStartä Taq DNA Polymerase D9307
JumpStartä Taq DNA Polymerase without MgCl2 D4184
JumpStartä REDTaqä DNA Polymerase D8187
JumpStartä Taq ReadyMixä P2893
JumpStartä REDTaqä ReadyMixä PCR Reaction Mix P0982
JumpStartä REDTaqä ReadyMixä Reaction Mix for High Throughput PCR P1107
JumpStartä AccuTaqä LA DNA Polymerase Mix D5809
JumpStartä REDAccuTaqä LA DNA Polymerase Mix D1313
JumpStartä KlenTaq® LA DNA Polymerase D0816
JumpStartä Taq Antibody A7721
Colony PCR Kit COL-PCR
back to top
Technical Data
Market Leading Performance of JumpStart Taq Enzymes
Inactivated by a Taq-Directed Antibody, JumpStart Taq Provides Increased Sensitivity, Specificity, and Yield
Sensitivity, Specificity, Yield
Ten nanograms (even lanes) or 100 nanograms (odd lanes) total human genomic DNA was amplified with primers targeted to a 5kb section of b-globin. Each PCR was performed according to supplier's recommendations.
Greater Specificity & Increased Yield with JumpStart Taq
JumpStart Taq Delivers Greater Specificity JumpStart taq polymerase and other commercially available hot start enzymes were used to amplify a five kilobase fragment of b-globin. Taq enzymes were activated per supplier's recommendations.
Superior Amplification Over a Wide-Dynamic Range of Template
4 Nanograms
JumpStart REDTaq
Four nanograms of template DNA was amplified under standard PCR conditions (lanes 1-4) and Hot Start conditions using JumpStart REDTaq.
0.4 Nanograms
JumpStart REDTaq
Standard PCR conditions (lanes 1-4) and Hot Start conditions using JumpStart REDTaq were used to amplify 0.4 ng template DNA.
back to top
JumpStart REDTaq - Direct Load & Hot Start PCR . . . A Winning Combination!
REDTaq DNA Polymerase is a unique blend of Sigma's high quality Taq DNA Polymerase combined with an inert red dye. The unique benefits of REDTaq DNA Polymerase include quick recognition and confirmation of appropriate mixing. Additionally, the red dye serves as a tracking dye and migrates at approximately the same rate as a 125 bp fragment.
Confirmation of Taq Polymerase Addition
REDTaq Recognition Vial 1 contains no REDTaq, vial 2 has 2.5 units of REDTaq added to a 50 µl reaction volume, and vial 3 shows a REDTaq PCR reaction solution thoroughly mixed.
Loading Buffers or Tracking Dyes not Required
REDTaq Loading Samples may be added directly to an agarose gel after PCR without the addition of a loading buffer or tracking dye. The dye in REDTaq acts as a tracking dye migrating at approximately the same rate as a 125 bp fragment.
back to top
Exceptional Performance with JumpStart REDTaq ReadyMix
Hot Start PCR in the Convenience of a ReadyMix
Superior Performance with the Convenience of a ReadyMix 200 ng Lambda phage DNA was amplified with Sigma's JumpStart REDTaq ReadyMix (odd numbered lanes) and Competitor I's Direct Load ReadyMix (even numbered lanes). Taq was activated per the supplier's recommendations.
back to top
back to PCR
Routine Amplification | Human Genomic DNA | Long & Accurate PCR Reagents | Hot Start PCR | RT-PCR | Quantitative PCR | Specialty Enzymes | Ultrapure Nucleotides | PCR Reaction Components | PCR Lab Equipment | DNA/RNA Molecular Weight Markers | Whole Genome Amplification