Molecular Biology
Automated Sequencing
Cloning and Expression
DNA and RNA Purification
Gene Expression Analysis
Molecular Biology Reagents
Nucleic Acid Detection
Nucleic Acid Electrophoresis
PCR
Product Lines
New PCR Products
Key Resources
Whole Genome Amplification
New Products
Neuroscience
Nutrition Research
Obesity Research
Peptides and Proteins
Plant Biotechnology
Proteomics and Protein Expr.
Stable Isotopes
Stem Cell Biology
Your Favorite Gene - Search
Life Science Innovations
PathFinder
 Capillary based Quantitative PCR
PCR
SYBR Green JumpStart Taq ReadyMix, Capillary formulation combines the advantages of a hot start enzyme, JumpStart taq, in a 2X concentrate ReadyMix specifically designed for use with capillary instruments, such as the Roche LightCycler® real–time thermal cycler.
  • Convenient – 2X concentrate ReadyMix designed specifically for use with capillary instruments.
  • Specific – JumpStart Taq antibody prevents non–specific product formation through it hot–start mechanism.
  • Optimized Formulation – Facilitate carry-over prevention from previous PCR with our specially formulated ReadyMix containing dUTP.
Product Name Technical Bulletin Product # 
SYBR Green JumpStart Taq ReadyMix for QPCR, Capillary formulation S1816
JumpStart Taq ReadyMix with dUTP D9191
Click Image to View Figure in its Entirety Figure 1. Efficient & Sensitive Amplification with Human Genomic DNA.
Back to Quantitative PCR home page.
Routine Amplification | Human Genomic DNA | Long & Accurate PCR Reagents | Hot Start PCR | RT-PCR | Quantitative PCR | Specialty Enzymes | Ultrapure Nucleotides | PCR Reaction Components | PCR Lab Equipment | DNA/RNA Molecular Weight Markers | Whole Genome Amplification